This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00210729_m1
See other PLA1A GE Assays ›
Gene Symbol:
PLA1A
Gene Name:
phospholipase A1 member A
Gene Aliases:
PS-PLA1, PSPLA1
Chromosome Location:
Chr.3: 119597848 - 119629811 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00210729_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLA1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001206960.1 NP_001193889.1 4 - 5 585 79
NM_001206961.1 NP_001193890.1 3 - 4 431 79
NM_001293225.1 NP_001280154.1 4 - 5 628 79
NM_015900.3 NP_056984.1 4 - 5 633 79
NR_120610.1 4 - 5 633 79
XM_017006572.1 XP_016862061.1 5 - 6 665 79
GenBank mRNA AF035268.1 - 4 - 5 598 79
AF035269.1 - 4 - 5 598 79
AK222705.1 - 4 - 5 79
AK301880.1 - 3 - 4 404 79
AK313519.1 - 4 - 5 596 79
BC035347.1 - 4 - 5 79
BC047703.2 - 4 - 5 537 6061929 79
BX647516.1 - 4 - 5 628 79
U37591.1 - 4 - 5 593 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300008484.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000611.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PLA1A

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
phosphatidylserine metabolic process
lipid catabolic process
phosphatidylcholine 1-acylhydrolase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000273371.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000472126.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000472126.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000472126.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000475963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000488919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000494440.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 ENST00000495992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206960.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001206961.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_001293225.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NM_015900.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008484.hg.1 NR_120610.1 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 157027 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0