This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00210856_m1
See other CALHM2 GE Assays ›
Gene Symbol:
CALHM2
Gene Name:
calcium homeostasis modulator 2
Gene Aliases:
FAM26B
Chromosome Location:
Chr.10: 103446785 - 103452419 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00210856_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CALHM2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_024552.1 3 - 4 1049 91
XM_006717884.3 XP_006717947.1 3 - 4 1672 91
XM_017016308.1 XP_016871797.1 3 - 4 1166 91
XM_017016309.1 XP_016871798.1 2 - 3 913 91
GenBank mRNA AF131810.1 - 3 - 4 910 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ion transmembrane transport
cation transmembrane transport
cation channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline