This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00212993_m1
See other BET1L GE Assays ›
Gene Symbol:
BET1L
Gene Name:
Bet1 golgi vesicular membrane trafficking protein like
Gene Aliases:
BET1L1, GOLIM3, GS15, HSPC197
Chromosome Location:
Chr.11: 202924 - 207422 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00212993_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BET1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001098787.1 NP_001092257.1 1 - 2 121 93
NM_016526.4 NP_057610.2 1 - 2 121 93
GenBank mRNA AF234160.1 - 1 - 2 122 93
AK022958.1 - 1 - 2 109 93
BC008971.1 - 1 - 2 112 2966914 93
BC032779.2 - 1 - 2 81 4938985 93
DA875317.1 - 1 - 2 121 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100013121.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11003513.hg.1

Additional Information:

This assay is in the 5'UTR region of the AF234160.1,AK022958.1,BC008971.1,BC032779.2,NM_001098787.1,NM_016526.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens 1700 Inventoried MicroArray Quality Control (MAQC) Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene BET1L

Panther Classification:

Molecular Function -

SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

ER to Golgi vesicle-mediated transport
protein transport
retrograde transport, endosome to Golgi
membrane fusion
regulation of retrograde vesicle-mediated transport, Golgi to ER
SNAP receptor activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11015401.hg.1 TC11003513.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11015401.hg.1 ENST00000325147.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11015401.hg.1 ENST00000332865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11015401.hg.1 ENST00000382762.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11015401.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11015401.hg.1 ENST00000529614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11015401.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11015401.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11015408.hg.1 TC11003513.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11015408.hg.1 ENST00000382762.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11015408.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11015408.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11015403.hg.1 TC11003513.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11015403.hg.1 ENST00000325147.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11015403.hg.1 ENST00000332865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11015403.hg.1 ENST00000382762.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11015403.hg.1 ENST00000529614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11015403.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11015403.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay PSR1100203161.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay PSR1100203161.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Clariom D Assay PSR1100203161.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay PSR1100203161.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100203161.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100203161.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay PSR1100203161.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay PSR1100203161.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay PSR1100203161.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Clariom D Assay PSR1100203161.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay PSR1100203161.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100203161.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100203161.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay PSR1100203161.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay PSR1100203161.hg.1 TC1100013121.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100203158.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay PSR1100203158.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Clariom D Assay PSR1100203158.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay PSR1100203158.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100203158.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100203158.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100203158.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay PSR1100203158.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay PSR1100203158.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay PSR1100203158.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Clariom D Assay PSR1100203158.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay PSR1100203158.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100203158.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100203158.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100203158.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay PSR1100203158.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay PSR1100203158.hg.1 TC1100013121.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay PSR1100203166.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay PSR1100203166.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay PSR1100203166.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay PSR1100203166.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay PSR1100203166.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay PSR1100203166.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay PSR1100203166.hg.1 TC1100013121.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 198548 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-tks2p:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0