This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00212994_m1
See other BET1L GE Assays ›
Gene Symbol:
BET1L
Gene Name:
Bet1 golgi vesicular membrane trafficking protein like
Gene Aliases:
BET1L1, GOLIM3, GS15, HSPC197
Chromosome Location:
Chr.11: 202924 - 207422 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00212994_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BET1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001098787.1 NP_001092257.1 2 - 3 213 67
NM_016526.4 NP_057610.2 2 - 3 213 67
GenBank mRNA AF234160.1 - 2 - 3 214 67
AK022958.1 - 2 - 3 201 67
BC008971.1 - 2 - 3 204 2966914 67
BC032779.2 - 2 - 3 173 4938985 67
DA875317.1 - 2 - 3 213 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100013121.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11003513.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried MicroArray Quality Control (MAQC) Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene BET1L

Panther Classification:

Molecular Function -

SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

ER to Golgi vesicle-mediated transport
protein transport
retrograde transport, endosome to Golgi
membrane fusion
regulation of retrograde vesicle-mediated transport, Golgi to ER
SNAP receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11015399.hg.1 TC11003513.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11015399.hg.1 ENST00000325147.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11015399.hg.1 ENST00000332865.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11015399.hg.1 ENST00000382762.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11015399.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11015399.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11015399.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11015398.hg.1 TC11003513.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11015398.hg.1 ENST00000325147.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11015398.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 ENST00000529614.6 Ensembl
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay TC1100013121.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay PSR1100203155.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay PSR1100203155.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100203155.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay PSR1100203155.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay PSR1100203155.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100203155.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay PSR1100203155.hg.1 TC1100013121.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100203156.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay PSR1100203156.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Clariom D Assay PSR1100203156.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay PSR1100203156.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100203156.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100203156.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay PSR1100203156.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay PSR1100203156.hg.1 ENST00000325147.13 Ensembl
Affymetrix Human Clariom D Assay PSR1100203156.hg.1 ENST00000332865.10 Ensembl
Affymetrix Human Clariom D Assay PSR1100203156.hg.1 ENST00000382762.7 Ensembl
Affymetrix Human Clariom D Assay PSR1100203156.hg.1 ENST00000479463.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100203156.hg.1 ENST00000486280.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100203156.hg.1 NM_001098787.1 Refseq
Affymetrix Human Clariom D Assay PSR1100203156.hg.1 NM_016526.4 Refseq
Affymetrix Human Clariom D Assay PSR1100203156.hg.1 TC1100013121.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0