This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00219347_m1
See other APBB1IP GE Assays ›
Gene Symbol:
APBB1IP
Gene Name:
amyloid beta precursor protein binding family B member 1 interacting protein
Gene Aliases:
INAG1, PREL1, RARP1, RIAM
Chromosome Location:
Chr.10: 26438324 - 26567879 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00219347_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APBB1IP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019043.3 NP_061916.3 2 - 3 319 129
XM_011519514.2 XP_011517816.1 2 - 3 332 129
GenBank mRNA AK000812.1 - 2 - 3 278 129
AK303597.1 - 2 - 3 879 129
AK308916.1 - 2 - 3 300 129
AY152730.1 - 1 - 2 85 129
BC035636.1 - 2 - 3 231 129
BC054516.1 - 1 - 2 97 5172946 129
BM150270.1 - 2 - 3 320 129
BM556115.1 - 286 129

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000007072.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000181.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK000812.1,AK303597.1,AY152730.1,BC035636.1,BC054516.1,NM_019043.3,XM_011519514.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene APBB1IP

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
signal transduction
positive regulation of cell adhesion
protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10002158.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10002158.hg.1 ENST00000376236.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10002158.hg.1 ENST00000445780.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10002158.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10002158.hg.1 TC10000181.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000362863.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000493857.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000493857.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000493857.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay PSR1000148235.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000148235.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000148235.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay PSR1000148235.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000148235.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000148235.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay PSR1000148235.hg.1 TC1000007072.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000362863.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000493857.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000493857.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000493857.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 132885 - -
ABI Human Genome Survey Microarray Version 2.0 202785 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0