This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00221260_m1
See other GPR107 GE Assays ›
Gene Symbol:
GPR107
Gene Name:
G protein-coupled receptor 107
Gene Aliases:
GCDRP, LUSTR1, bA138E2.2
Chromosome Location:
Chr.9: 130053706 - 130140169 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00221260_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR107
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK026244.1 - 9 - 10 876 68
AK225456.1 - 9 - 10 876 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Homo sapiens 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 134678 - -
ABI Human Genome Survey Microarray Version 2.0 165057 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0