This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00228981_m1
See other STARD5 GE Assays ›
Gene Symbol:
STARD5
Gene Name:
StAR related lipid transfer domain containing 5
Gene Aliases:
-
Chromosome Location:
Chr.15: 81312666 - 81324183 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00228981_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STARD5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_135013.1 1 - 2 188 81
GenBank mRNA AK026352.1 - 1 - 2 146 81
AK225479.1 - 1 - 2 146 81
AL137657.1 - 1 - 2 188 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Homo sapiens 1700 Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

C21-steroid hormone biosynthetic process
lipid transport
cholesterol binding
bile acid binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000325346.6 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000559913.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560156.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 ENST00000560723.5 Ensembl
Affymetrix Human Clariom D Assay TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay TC1500010909.hg.1 NM_181900.2 Refseq
Affymetrix Human Clariom D Assay TC1500010909.hg.1 NM_181900.2 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 210466 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4d4pq:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0