This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00242338_m1
See other APOBEC1 GE Assays ›
Gene Symbol:
APOBEC1
Gene Name:
apolipoprotein B mRNA editing enzyme catalytic subunit 1
Gene Aliases:
APOBEC-1, BEDP, CDAR1, HEPR
Chromosome Location:
Chr.12: 7649400 - 7670599 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00242338_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APOBEC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001304566.1 NP_001291495.1 2 - 3 63 69
NM_001644.4 NP_001635.2 2 - 3 70 69
GenBank mRNA BC096158.1 - 2 - 3 60 69
BC101972.1 - 2 - 3 57 69
BC101973.1 - 2 - 3 60 69
L25877.1 - 2 - 3 61 69
L26234.1 - 2 - 3 63 69
U72891.1 - 2 - 3 61 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001163.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene APOBEC1

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

RNA processing
mRNA processing
lipid metabolic process
response to osmotic stress
cytidine deamination
response to zinc ion
response to gamma radiation
cytidine to uridine editing
mRNA modification
cellular response to insulin stimulus
regulation of cell proliferation
lipoprotein biosynthetic process
response to drug
lipoprotein transport
positive regulation of catalytic activity
response to ethanol
mRNA stabilization
response to calcium ion
defense response to virus
DNA cytosine deamination
DNA demethylation
negative regulation of methylation-dependent chromatin silencing
positive regulation of mRNA modification
RNA binding
mRNA binding
cytidine deaminase activity
cytosine deaminase activity
protein binding
enzyme activator activity
zinc ion binding
AU-rich element binding
protein domain specific binding
ribonucleoprotein complex binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR12015485.hg.1 ENST00000229304.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12015485.hg.1 NM_001644.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12015485.hg.1 TC12001163.hg.1 HTA TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 154821 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0