This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00242501_m1
See other E2F6 GE Assays ›
Gene Symbol:
E2F6
Gene Name:
E2F transcription factor 6
Gene Aliases:
E2F-6
Chromosome Location:
Chr.2: 11444375 - 11466177 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00242501_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: E2F6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_198256.3 NP_937987.2 2 - 3 461 109
NR_103490.1 2 - 3 461 109
XM_017003547.1 XP_016859036.1 2 - 3 445 109
GenBank mRNA AF041381.1 - 2 - 3 147 109
AF059292.1 - 2 - 3 163 109
AK223159.1 - 2 - 3 134 109
AK223407.1 - 2 - 3 440 109
AK290413.1 - 2 - 3 433 109
AK293504.1 - 2 - 3 433 109
AU132447.1 - 2 - 3 435 109
BC008348.1 - 2 - 3 420 3535798 109
BC107740.1 - 2 - 3 378 6494695 109
KU178035.1 - 2 - 3 163 109

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001579.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Human Transcription Factors (non-Hox) Human p53 Signaling HUMAN FACTORS PROMOTING CARDIOGENESIS IN VERTEBRATES 1700 Inventoried Human_p53_Signaling Human_Transcription_Factors_(non-Hox) Human_Factors_Promoting_Cardiogenesis_in_Vertebrates Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene E2F6

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription involved in G1/S transition of mitotic cell cycle
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02025501.hg.1 TC02001579.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02025501.hg.1 ENST00000307236.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02025501.hg.1 ENST00000362009.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02025501.hg.1 ENST00000381525.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02025501.hg.1 ENST00000542100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02025501.hg.1 ENST00000546212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02025501.hg.1 NM_198256.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02025500.hg.1 TC02001579.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02025500.hg.1 ENST00000307236.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02025500.hg.1 ENST00000362009.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02025500.hg.1 ENST00000381525.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02025500.hg.1 ENST00000542100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02025500.hg.1 ENST00000546212.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02025500.hg.1 NM_198256.3 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 193661 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0