This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00245542_m1
See other GPR18 GE Assays ›
Gene Symbol:
GPR18
Gene Name:
G protein-coupled receptor 18
Gene Aliases:
-
Chromosome Location:
Chr.13: 99254713 - 99262498 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00245542_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001098200.1 NP_001091670.1 1 - 2 270 91
XM_006719946.3 XP_006720009.1 2 - 3 1272 91
GenBank mRNA AF339781.1 - 1 - 2 218 91
BC008569.1 - 1 - 2 229 4338810 91
BC066927.1 - 2 - 3 518 4373724 91
BI770379.1 - 1 - 2 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300009597.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000828.hg.1

Additional Information:

This assay is in the 5'UTR region of the BC008569.1,BC066927.1,NM_001098200.1,XM_006719946.3 transcripts.

Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR13009335.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13009335.hg.1 ENST00000397470.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13009335.hg.1 ENST00000397473.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13009335.hg.1 NM_001098200.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13009335.hg.1 NM_005292.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13009335.hg.1 TC13000828.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000397470.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000397470.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000397470.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000397470.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000397470.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000397470.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000416594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000416594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000416594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 ENST00000416594.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 NM_001098200.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 NM_001098200.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 NM_001098200.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 NM_001098200.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 NM_001098200.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 NM_001098200.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 NM_005292.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 NM_005292.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 NM_005292.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 NM_005292.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 NM_005292.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 NM_005292.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009597.hg.1 NM_005292.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13009340.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13009340.hg.1 ENST00000397470.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13009340.hg.1 ENST00000397473.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13009340.hg.1 NM_001098200.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13009340.hg.1 NM_005292.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13009340.hg.1 TC13000828.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR13009336.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13009336.hg.1 ENST00000397470.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13009336.hg.1 ENST00000397473.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13009336.hg.1 NM_001098200.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13009336.hg.1 NM_005292.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13009336.hg.1 TC13000828.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR13009334.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13009334.hg.1 ENST00000397470.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13009334.hg.1 ENST00000397473.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13009334.hg.1 NM_001098200.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13009334.hg.1 NM_005292.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13009334.hg.1 TC13000828.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR13009338.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13009338.hg.1 ENST00000397470.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13009338.hg.1 ENST00000397473.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13009338.hg.1 NM_001098200.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13009338.hg.1 NM_005292.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13009338.hg.1 TC13000828.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1300164211.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Clariom D Assay PSR1300164211.hg.1 ENST00000397470.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300164211.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Clariom D Assay PSR1300164211.hg.1 ENST00000416594.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300164211.hg.1 NM_001098200.1 Refseq
Affymetrix Human Clariom D Assay PSR1300164211.hg.1 NM_005292.3 Refseq
Affymetrix Human Clariom D Assay PSR1300164211.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Clariom D Assay PSR1300164211.hg.1 ENST00000397470.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300164211.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Clariom D Assay PSR1300164211.hg.1 ENST00000416594.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300164211.hg.1 NM_001098200.1 Refseq
Affymetrix Human Clariom D Assay PSR1300164211.hg.1 NM_005292.3 Refseq
Affymetrix Human Clariom D Assay PSR1300164211.hg.1 TC1300009597.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1300164210.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Clariom D Assay PSR1300164210.hg.1 ENST00000397470.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300164210.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Clariom D Assay PSR1300164210.hg.1 NM_001098200.1 Refseq
Affymetrix Human Clariom D Assay PSR1300164210.hg.1 NM_005292.3 Refseq
Affymetrix Human Clariom D Assay PSR1300164210.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Clariom D Assay PSR1300164210.hg.1 ENST00000397470.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300164210.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Clariom D Assay PSR1300164210.hg.1 NM_001098200.1 Refseq
Affymetrix Human Clariom D Assay PSR1300164210.hg.1 NM_005292.3 Refseq
Affymetrix Human Clariom D Assay PSR1300164210.hg.1 TC1300009597.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1300164216.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Clariom D Assay PSR1300164216.hg.1 ENST00000397470.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300164216.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Clariom D Assay PSR1300164216.hg.1 NM_001098200.1 Refseq
Affymetrix Human Clariom D Assay PSR1300164216.hg.1 NM_005292.3 Refseq
Affymetrix Human Clariom D Assay PSR1300164216.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Clariom D Assay PSR1300164216.hg.1 ENST00000397470.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300164216.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Clariom D Assay PSR1300164216.hg.1 NM_001098200.1 Refseq
Affymetrix Human Clariom D Assay PSR1300164216.hg.1 NM_005292.3 Refseq
Affymetrix Human Clariom D Assay PSR1300164216.hg.1 TC1300009597.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1300164217.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Clariom D Assay PSR1300164217.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Clariom D Assay PSR1300164217.hg.1 TC1300009597.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1300164218.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Clariom D Assay PSR1300164218.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Clariom D Assay PSR1300164218.hg.1 TC1300009597.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1300164212.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Clariom D Assay PSR1300164212.hg.1 ENST00000397470.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300164212.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Clariom D Assay PSR1300164212.hg.1 ENST00000416594.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300164212.hg.1 NM_001098200.1 Refseq
Affymetrix Human Clariom D Assay PSR1300164212.hg.1 NM_005292.3 Refseq
Affymetrix Human Clariom D Assay PSR1300164212.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Clariom D Assay PSR1300164212.hg.1 ENST00000397470.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300164212.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Clariom D Assay PSR1300164212.hg.1 ENST00000416594.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300164212.hg.1 NM_001098200.1 Refseq
Affymetrix Human Clariom D Assay PSR1300164212.hg.1 NM_005292.3 Refseq
Affymetrix Human Clariom D Assay PSR1300164212.hg.1 TC1300009597.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000397470.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000397470.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000397470.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000397470.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000397470.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000397470.4 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000416594.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000416594.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000416594.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 ENST00000416594.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009597.hg.1 NM_001098200.1 Refseq
Affymetrix Human Clariom D Assay TC1300009597.hg.1 NM_001098200.1 Refseq
Affymetrix Human Clariom D Assay TC1300009597.hg.1 NM_001098200.1 Refseq
Affymetrix Human Clariom D Assay TC1300009597.hg.1 NM_001098200.1 Refseq
Affymetrix Human Clariom D Assay TC1300009597.hg.1 NM_001098200.1 Refseq
Affymetrix Human Clariom D Assay TC1300009597.hg.1 NM_001098200.1 Refseq
Affymetrix Human Clariom D Assay TC1300009597.hg.1 NM_005292.3 Refseq
Affymetrix Human Clariom D Assay TC1300009597.hg.1 NM_005292.3 Refseq
Affymetrix Human Clariom D Assay TC1300009597.hg.1 NM_005292.3 Refseq
Affymetrix Human Clariom D Assay TC1300009597.hg.1 NM_005292.3 Refseq
Affymetrix Human Clariom D Assay TC1300009597.hg.1 NM_005292.3 Refseq
Affymetrix Human Clariom D Assay TC1300009597.hg.1 NM_005292.3 Refseq
Affymetrix Human Clariom D Assay TC1300009597.hg.1 NM_005292.3 Refseq
Affymetrix Human Clariom D Assay PSR1300164214.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Clariom D Assay PSR1300164214.hg.1 ENST00000397470.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300164214.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Clariom D Assay PSR1300164214.hg.1 ENST00000416594.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300164214.hg.1 NM_001098200.1 Refseq
Affymetrix Human Clariom D Assay PSR1300164214.hg.1 NM_005292.3 Refseq
Affymetrix Human Clariom D Assay PSR1300164214.hg.1 ENST00000340807.3 Ensembl
Affymetrix Human Clariom D Assay PSR1300164214.hg.1 ENST00000397470.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300164214.hg.1 ENST00000397473.6 Ensembl
Affymetrix Human Clariom D Assay PSR1300164214.hg.1 ENST00000416594.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300164214.hg.1 NM_001098200.1 Refseq
Affymetrix Human Clariom D Assay PSR1300164214.hg.1 NM_005292.3 Refseq
Affymetrix Human Clariom D Assay PSR1300164214.hg.1 TC1300009597.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 118386 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0