This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00245978_m1
See other CD200 GE Assays ›
Gene Symbol:
CD200
Gene Name:
CD200 molecule
Gene Aliases:
MOX1, MOX2, MRC, OX-2
Chromosome Location:
Chr.3: 112332224 - 112362812 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00245978_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CD200
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001318826.1 NP_001305755.1 1 - 2 282 78
NM_005944.6 NP_005935.4 1 - 2 282 78
XM_005247482.2 XP_005247539.1 1 - 2 161 78
XM_017006457.1 XP_016861946.1 1 - 2 161 78
GenBank mRNA AF063591.1 - 1 - 2 73 78
AK074972.1 - 1 - 2 47 78
AL832529.1 - 1 - 2 67 78
BC022522.1 - 1 - 2 75 4794050 78
BC031103.1 - 1 - 2 51 5301702 78
DA544134.1 - 1 - 2 160 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300013859.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000572.hg.1

Additional Information:

This assay is in the 5'UTR region of the AF063591.1,AK074972.1,BC022522.1,BC031103.1,NM_001318826.1,NM_005944.6,XM_005247482.2,XM_017006457.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CD200

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
cell recognition
negative regulation of macrophage activation
regulation of immune response
receptor activity
receptor binding
protein binding
protein homodimerization activity
cell adhesion molecule binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03010384.hg.1 ENST00000315711.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03010384.hg.1 ENST00000383681.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03010384.hg.1 ENST00000473539.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03010384.hg.1 NM_001004196.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03010384.hg.1 NM_005944.6 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03010384.hg.1 TC03000572.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000315711.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000315711.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000315711.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000315711.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000315711.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000315711.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000315711.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000383681.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000383681.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000383681.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000383681.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000383681.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000383681.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000473539.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000473539.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000473539.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000473539.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000473539.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000473539.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000473539.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000478595.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000478595.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000478595.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000478595.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000478595.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000478595.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000498096.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000498096.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000498096.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000498096.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000498096.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000498096.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000607516.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000607516.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000607516.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000607516.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 ENST00000607516.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_001004196.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_001004196.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_001004196.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_001004196.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_001004196.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_001004196.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_001004196.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_001004196.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_001004196.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_005944.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_005944.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_005944.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_005944.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_005944.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_005944.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_005944.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013859.hg.1 NM_005944.6 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03010382.hg.1 NM_001004196.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03010382.hg.1 NM_005944.6 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03010382.hg.1 TC03000572.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03010387.hg.1 ENST00000315711.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03010387.hg.1 ENST00000383681.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03010387.hg.1 ENST00000473539.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03010387.hg.1 NM_001004196.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03010387.hg.1 NM_005944.6 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03010387.hg.1 TC03000572.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03010386.hg.1 ENST00000315711.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03010386.hg.1 ENST00000473539.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03010386.hg.1 NM_001004196.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03010386.hg.1 NM_005944.6 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03010386.hg.1 TC03000572.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0300205046.hg.1 ENST00000498096.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300205046.hg.1 ENST00000607516.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300205046.hg.1 NM_001004196.3 Refseq
Affymetrix Human Clariom D Assay PSR0300205046.hg.1 NM_005944.6 Refseq
Affymetrix Human Clariom D Assay PSR0300205046.hg.1 ENST00000498096.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300205046.hg.1 ENST00000607516.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300205046.hg.1 NM_001004196.3 Refseq
Affymetrix Human Clariom D Assay PSR0300205046.hg.1 NM_005944.6 Refseq
Affymetrix Human Clariom D Assay PSR0300205046.hg.1 TC0300013859.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300205053.hg.1 ENST00000315711.12 Ensembl
Affymetrix Human Clariom D Assay PSR0300205053.hg.1 ENST00000473539.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300205053.hg.1 ENST00000498096.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300205053.hg.1 NM_001004196.3 Refseq
Affymetrix Human Clariom D Assay PSR0300205053.hg.1 NM_005944.6 Refseq
Affymetrix Human Clariom D Assay PSR0300205053.hg.1 ENST00000315711.12 Ensembl
Affymetrix Human Clariom D Assay PSR0300205053.hg.1 ENST00000473539.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300205053.hg.1 ENST00000498096.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300205053.hg.1 NM_001004196.3 Refseq
Affymetrix Human Clariom D Assay PSR0300205053.hg.1 NM_005944.6 Refseq
Affymetrix Human Clariom D Assay PSR0300205053.hg.1 TC0300013859.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000315711.12 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000315711.12 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000315711.12 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000315711.12 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000315711.12 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000315711.12 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000315711.12 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000383681.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000383681.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000383681.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000383681.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000383681.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000383681.7 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000473539.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000473539.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000473539.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000473539.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000473539.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000473539.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000473539.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000478595.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000478595.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000478595.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000478595.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000478595.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000478595.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000498096.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000498096.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000498096.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000498096.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000498096.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000498096.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000607516.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000607516.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000607516.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000607516.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 ENST00000607516.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_001004196.3 Refseq
Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_001004196.3 Refseq
Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_001004196.3 Refseq
Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_001004196.3 Refseq
Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_001004196.3 Refseq
Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_001004196.3 Refseq
Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_001004196.3 Refseq
Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_001004196.3 Refseq
Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_001004196.3 Refseq
Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_005944.6 Refseq
Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_005944.6 Refseq
Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_005944.6 Refseq
Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_005944.6 Refseq
Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_005944.6 Refseq
Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_005944.6 Refseq
Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_005944.6 Refseq
Affymetrix Human Clariom D Assay TC0300013859.hg.1 NM_005944.6 Refseq
Affymetrix Human Clariom D Assay PSR0300205048.hg.1 ENST00000315711.12 Ensembl
Affymetrix Human Clariom D Assay PSR0300205048.hg.1 ENST00000383681.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300205048.hg.1 ENST00000473539.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300205048.hg.1 ENST00000478595.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300205048.hg.1 ENST00000498096.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300205048.hg.1 ENST00000607516.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300205048.hg.1 NM_001004196.3 Refseq
Affymetrix Human Clariom D Assay PSR0300205048.hg.1 NM_005944.6 Refseq
Affymetrix Human Clariom D Assay PSR0300205048.hg.1 ENST00000315711.12 Ensembl
Affymetrix Human Clariom D Assay PSR0300205048.hg.1 ENST00000383681.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300205048.hg.1 ENST00000473539.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300205048.hg.1 ENST00000478595.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300205048.hg.1 ENST00000498096.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300205048.hg.1 ENST00000607516.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300205048.hg.1 NM_001004196.3 Refseq
Affymetrix Human Clariom D Assay PSR0300205048.hg.1 NM_005944.6 Refseq
Affymetrix Human Clariom D Assay PSR0300205048.hg.1 TC0300013859.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 105368 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline