This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00251998_m1
See other GPR162 GE Assays ›
Gene Symbol:
GPR162
Gene Name:
G protein-coupled receptor 162
Gene Aliases:
A-2, GRCA
Chromosome Location:
Chr.12: 6821797 - 6827418 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00251998_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR162
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019858.1 NP_062832.1 2 - 3 1403 76
GenBank mRNA AB209836.1 - 1 - 2 810 76
U47928.1 - 2 - 3 1403 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200012587.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12003231.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR162

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000382315.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000382315.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000382315.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000382315.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000382315.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000382315.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000382315.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000428545.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000428545.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000428545.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000428545.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000428545.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000428545.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000428545.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000428545.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000428545.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000428545.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000541431.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000541431.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000541431.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000541431.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000542330.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000542330.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 ENST00000542330.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 NM_014449.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 NM_014449.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 NM_014449.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 NM_014449.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 NM_014449.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 NM_014449.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 NM_014449.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 NM_014449.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 NM_014449.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12001072.hg.1 TC12003231.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12001072.hg.1 ENST00000311268.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12001072.hg.1 NM_019858.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12001070.hg.1 TC12003231.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12001070.hg.1 ENST00000311268.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12001070.hg.1 ENST00000541431.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12001070.hg.1 NM_019858.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12001071.hg.1 TC12003231.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12001071.hg.1 ENST00000311268.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12001071.hg.1 NM_019858.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12001073.hg.1 TC12003231.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12001073.hg.1 ENST00000311268.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12001073.hg.1 NM_019858.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12001069.hg.1 TC12003231.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12001069.hg.1 ENST00000311268.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12001069.hg.1 NM_019858.1 Refseq
Affymetrix Human Clariom D Assay PSR1200198922.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200198922.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200198922.hg.1 NM_019858.1 Refseq
Affymetrix Human Clariom D Assay PSR1200198922.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200198922.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200198922.hg.1 NM_019858.1 Refseq
Affymetrix Human Clariom D Assay PSR1200198922.hg.1 TC1200012587.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200198921.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200198921.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200198921.hg.1 ENST00000541431.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200198921.hg.1 NM_019858.1 Refseq
Affymetrix Human Clariom D Assay PSR1200198921.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200198921.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200198921.hg.1 ENST00000541431.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200198921.hg.1 NM_019858.1 Refseq
Affymetrix Human Clariom D Assay PSR1200198921.hg.1 TC1200012587.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200198919.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200198919.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200198919.hg.1 NM_019858.1 Refseq
Affymetrix Human Clariom D Assay PSR1200198919.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200198919.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200198919.hg.1 NM_019858.1 Refseq
Affymetrix Human Clariom D Assay PSR1200198919.hg.1 TC1200012587.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200198920.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200198920.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200198920.hg.1 NM_019858.1 Refseq
Affymetrix Human Clariom D Assay PSR1200198920.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200198920.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200198920.hg.1 NM_019858.1 Refseq
Affymetrix Human Clariom D Assay PSR1200198920.hg.1 TC1200012587.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000311268.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000382315.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000382315.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000382315.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000382315.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000382315.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000382315.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000382315.7 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000428545.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000428545.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000428545.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000428545.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000428545.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000428545.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000428545.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000428545.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000428545.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000428545.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000535220.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000541431.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000541431.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000541431.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000541431.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000542330.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000542330.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 ENST00000542330.1 Ensembl
Affymetrix Human Clariom D Assay TC1200012587.hg.1 NM_014449.1 Refseq
Affymetrix Human Clariom D Assay TC1200012587.hg.1 NM_014449.1 Refseq
Affymetrix Human Clariom D Assay TC1200012587.hg.1 NM_014449.1 Refseq
Affymetrix Human Clariom D Assay TC1200012587.hg.1 NM_014449.1 Refseq
Affymetrix Human Clariom D Assay TC1200012587.hg.1 NM_014449.1 Refseq
Affymetrix Human Clariom D Assay TC1200012587.hg.1 NM_014449.1 Refseq
Affymetrix Human Clariom D Assay TC1200012587.hg.1 NM_014449.1 Refseq
Affymetrix Human Clariom D Assay TC1200012587.hg.1 NM_014449.1 Refseq
Affymetrix Human Clariom D Assay TC1200012587.hg.1 NM_014449.1 Refseq
Affymetrix Human Clariom D Assay TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Clariom D Assay TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Clariom D Assay TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Clariom D Assay TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Clariom D Assay TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Clariom D Assay TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Clariom D Assay TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Clariom D Assay TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Clariom D Assay TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Clariom D Assay TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Clariom D Assay TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Clariom D Assay TC1200012587.hg.1 NM_019858.1 Refseq
Affymetrix Human Clariom D Assay TC1200012587.hg.1 NM_019858.1 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 125615 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0