This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00254277_m1
See other NAA35 GE Assays ›
Gene Symbol:
NAA35
Gene Name:
N(alpha)-acetyltransferase 35, NatC auxiliary subunit
Gene Aliases:
EGAP, MAK10, MAK10P, bA379P1.1
Chromosome Location:
Chr.9: 85941142 - 86026355 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00254277_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAA35
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001321881.1 NP_001308810.1 15 - 16 1422 88
NM_001321882.1 NP_001308811.1 15 - 16 1640 88
NM_024635.3 NP_078911.3 15 - 16 1426 88
XM_005252127.3 XP_005252184.1 15 - 16 1497 88
XM_011518903.2 XP_011517205.1 12 - 13 1229 88
XM_011518904.2 XP_011517206.1 9 - 10 915 88
XM_017015012.1 XP_016870501.1 16 - 17 1485 88
GenBank mRNA AK025266.1 - 15 - 16 1422 88
AK225237.1 - 15 - 16 1422 88
AK292239.1 - 15 - 16 1426 88
BC117427.1 - 15 - 16 1347 40125978 88
BC117429.1 - 15 - 16 1343 40125980 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900007775.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09000391.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NAA35

Panther Classification:

Molecular Function -

acetyltransferase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of apoptotic process
smooth muscle cell proliferation
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09004198.hg.1 TC09000391.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09004198.hg.1 ENST00000361671.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09004198.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000376040.2 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 ENST00000416045.4 Ensembl
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay TC0900007775.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay PSR0900152551.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay PSR0900152551.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay PSR0900152551.hg.1 ENST00000361671.9 Ensembl
Affymetrix Human Clariom D Assay PSR0900152551.hg.1 NM_024635.3 Refseq
Affymetrix Human Clariom D Assay PSR0900152551.hg.1 TC0900007775.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-fmmxr:80/100.66.76.202:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0