This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00254433_m1
See other GPSM3 GE Assays ›
Gene Symbol:
GPSM3
Gene Name:
G-protein signaling modulator 3
Gene Aliases:
AGS4, C6orf9, G18, G18.1a, G18.1b, G18.2, NG1
Chromosome Location:
Chr.6: 32190766 - 32195523 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00254433_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPSM3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022107.2 NP_071390.1 1 - 2 97 88
GenBank mRNA AJ243937.1 - 1 - 2 97 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06001563.hg.1, TC6_apd_hap1000097.hg.1, TC6_cox_hap2000189.hg.1, TC6_mann_hap4000154.hg.1, TC6_mcf_hap5000164.hg.1, TC6_qbl_hap6000178.hg.1, TC6_ssto_hap7000158.hg.1

Additional Information:

This assay is in the 5'UTR region of the AJ243937.1,NM_022107.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPSM3

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of leukocyte chemotaxis
biological_process
regulation of catalytic activity
positive regulation of cytokine production involved in inflammatory response
molecular_function
protein binding
GTPase regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR06018469.hg.1 TC06001563.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06018469.hg.1 ENST00000375043.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06018469.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06018470.hg.1 TC06001563.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06018470.hg.1 ENST00000375043.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06018470.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06018471.hg.1 TC06001563.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06018471.hg.1 ENST00000375043.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06018471.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_apd_hap1001566.hg.1 TC6_apd_hap1000097.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_apd_hap1001566.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003694.hg.1 TC6_cox_hap2000189.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003694.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4002830.hg.1 TC6_mann_hap4000154.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4002830.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5003368.hg.1 TC6_mcf_hap5000164.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5003368.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_apd_hap1001565.hg.1 TC6_apd_hap1000097.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_apd_hap1001565.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5003366.hg.1 TC6_mcf_hap5000164.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5003366.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003527.hg.1 TC6_qbl_hap6000178.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003527.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4002829.hg.1 TC6_mann_hap4000154.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4002829.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5003367.hg.1 TC6_mcf_hap5000164.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5003367.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003529.hg.1 TC6_qbl_hap6000178.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003529.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_apd_hap1001567.hg.1 TC6_apd_hap1000097.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_apd_hap1001567.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003692.hg.1 TC6_cox_hap2000189.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003692.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003528.hg.1 TC6_qbl_hap6000178.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003528.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7003258.hg.1 TC6_ssto_hap7000158.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7003258.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003693.hg.1 TC6_cox_hap2000189.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003693.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4002831.hg.1 TC6_mann_hap4000154.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4002831.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7003259.hg.1 TC6_ssto_hap7000158.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7003259.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7003260.hg.1 TC6_ssto_hap7000158.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7003260.hg.1 NM_022107.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0