This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00254439_s1
See other OXCT2 GE Assays ›
Gene Symbol:
OXCT2 OXCT2P1
Gene Name:
3-oxoacid CoA-transferase 2
3-oxoacid CoA-transferase 2 pseudogene 1
Gene Aliases:
FKSG25, SCOTT
OXCT2P
Chromosome Location:
Chr.1: 39769525 - 39771348 on Build GRCh38
Chromosome Location:
Chr.1: 39514866 - 39516669 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs00254439_s1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OXCT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022120.1 NP_071403.1 1 233 85
GenBank mRNA AB050193.2 - 1 159 85
AK093089.1 - 1 207 85
AK314586.1 - 1 140 85
AY013700.1 - 1 233 85
BC030015.1 - 1 152 85
Gene Symbol: OXCT2P1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_132962.1 1 230 85
GenBank mRNA BC072388.1 - 1 230 5588771 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100007910.hg.1, TC0100013805.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01002548.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene OXCT2
Click here to find siRNAs for Gene OXCT2P1

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

ketone body catabolic process
3-oxoacid CoA-transferase activity

MicroArray Probes:

Agilent Affymetrix ArrayStar ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Human Whole Genome ChIP Tiling Array A_17_P00153074 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00153080 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00153073 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20177317 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00153071 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00153084 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00153078 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00153070 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20177327 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00153072 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00153082 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00153079 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P00153076 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P20177335 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_23_P137896 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P85123 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P137896 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 30799 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 38170 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 25425 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_23_P137896 - -
SurePrint G3 Human GE Microarray G4851A A_23_P137896 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00153072 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00153076 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00153069 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00153073 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20177310 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15051770 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00153070 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00153067 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00153078 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20177327 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20177317 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00153071 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20177327 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00153076 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00153067 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00153069 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00153071 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00153078 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00153072 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20177310 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00153070 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20177317 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15051770 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00153073 - -
Whole Human Genome Oligo Microarray G4112A A_23_P137896 - -
Whole Human Genome Oligo Microarray G4112A A_24_P85123 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P00153069 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P00153072 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P00153075 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P00153067 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P00153074 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P00153068 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15051740 - -
Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P137896 - -
Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P85123 - -
Agilent Human 1B Microarray G4111A A_32_P364829 - -
Agilent Human 1B Microarray G4111A A_32_P124828 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00153086 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00153068 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00153080 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00153067 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00153072 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00153075 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00153078 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00153084 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00153079 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P15051740 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P00153074 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array 7915260 - -
Affymetrix Human Gene 1.0 ST Array 7900353 - -
Affymetrix Human Gene 1.0 ST Array 7915258 - -
Affymetrix Human Gene 1.0 ST Array 7915259 - -
Affymetrix Human Gene 1.1 ST Array 7900352 - -
Affymetrix Human Gene 1.1 ST Array 7915257 - -
Affymetrix HT HG-U133+ PM Array Plate 233301_PM_at - -
Affymetrix HT HG-U133+ PM Array Plate 220256_PM_s_at - -
Affymetrix Human Genome U219 Array 11758305_s_at - -
Affymetrix Human Genome U219 Array 11757081_s_at - -
Affymetrix Human Genome U219 Array 11758305_PM_s_at - -
Affymetrix Human Genome U219 Array 11757081_PM_s_at - -
Affymetrix Human Transcriptome Array 2.0 TC0100007910.hg.1 ENST00000447263.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01039625.hg.1 TC01002548.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01039625.hg.1 ENST00000327582.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01039625.hg.1 ENST00000542949.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01039625.hg.1 NM_022120.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013805.hg.1 ENST00000327582.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013805.hg.1 ENST00000327582.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013805.hg.1 ENST00000327582.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013805.hg.1 NM_022120.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013805.hg.1 NM_022120.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013805.hg.1 NM_022120.1 Refseq
Affymetrix Human Clariom D Assay PSR0100203240.hg.1 ENST00000327582.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100203240.hg.1 NM_022120.1 Refseq
Affymetrix Human Clariom D Assay PSR0100203240.hg.1 ENST00000327582.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100203240.hg.1 NM_022120.1 Refseq
Affymetrix Human Clariom D Assay PSR0100203240.hg.1 TC0100013805.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100007910.hg.1 ENST00000447263.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100203241.hg.1 ENST00000327582.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100203241.hg.1 NM_022120.1 Refseq
Affymetrix Human Clariom D Assay PSR0100203241.hg.1 ENST00000327582.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100203241.hg.1 NM_022120.1 Refseq
Affymetrix Human Clariom D Assay PSR0100203241.hg.1 TC0100013805.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100155676.hg.1 TC0100007910.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100155674.hg.1 TC0100007910.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100155675.hg.1 ENST00000447263.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100155675.hg.1 ENST00000447263.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100155675.hg.1 TC0100007910.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100013805.hg.1 ENST00000327582.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013805.hg.1 ENST00000327582.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013805.hg.1 ENST00000327582.5 Ensembl
Affymetrix Human Clariom D Assay TC0100013805.hg.1 NM_022120.1 Refseq
Affymetrix Human Clariom D Assay TC0100013805.hg.1 NM_022120.1 Refseq
Affymetrix Human Clariom D Assay TC0100013805.hg.1 NM_022120.1 Refseq
Human Genome U133 Plus 2.0 Array 220256_s_at - -
Human Genome U133 Plus 2.0 Array 233301_PM_at - -
Human Genome U133 Plus 2.0 Array 220256_PM_s_at - -
Human Genome U133 Plus 2.0 Array 233301_at - -
Human Genome U133A 2.0 Array 220256_PM_s_at - -
Human Genome U133A 2.0 Array 220256_s_at - -
AffymetrixPathwork Diagnostics Pathchip pw551695 - -
Affymetrix Human Genome U133 Plus 2.0 Array 220256_s_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 220256_s_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 233301_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 220256_s_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 220256_s_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 233301_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 233301_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 220256_s_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 233301_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 220256_s_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 233301_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 233301_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 220256_s_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 220256_s_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 233301_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 220256_s_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 233301_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 233301_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 220256_s_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 220256_s_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 233301_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 233301_at11 - -
Human Genome U95 Set 57726_PM_at - -
Human Genome U95 Set 57726_at - -
Human Genome U133 Set 220256_PM_s_at - -
Human Genome U133 Set 220256_s_at - -
Human Genome U133 Set 233301_PM_at - -
Human Genome U133 Set 233301_at - -
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A044039 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A044039 - -
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 152831 - -
ABI Human Genome Survey Microarray Version 2.0 179599 - -
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_1653730 BC072388.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_1653730 AY013700.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_1653730 BC030015.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_1653730 NM_022120.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_1653730 AB050193.2 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1653730 BC072388.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1653730 AY013700.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1653730 BC030015.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1653730 NM_022120.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1653730 AB050193.2 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1653730 BC072388.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1653730 AY013700.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1653730 BC030015.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1653730 NM_022120.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1653730 AB050193.2 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1653730 BC072388.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1653730 AY013700.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1653730 BC030015.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1653730 NM_022120.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1653730 AB050193.2 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1653730 BC072388.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1653730 AY013700.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1653730 BC030015.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1653730 NM_022120.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1653730 AB050193.2 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_11545840-S BC072388.1 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_11545840-S AY013700.1 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_11545840-S BC030015.1 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_11545840-S NM_022120.1 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_11545840-S AB050193.2 human_transcriptome
Illumina HumanRef-8 v2 Expression BeadChip GI_24641-S - -
Illumina human-6 v1.0 expression beadchip 0006290022 - -
Illumina humanRef-8 v1.0 expression beadchip 0006290022 - -
Illumina human-6 v2.0 expression beadchip ILMN_1653730 BC072388.1 human_transcriptome
Illumina human-6 v2.0 expression beadchip ILMN_1653730 AY013700.1 human_transcriptome
Illumina human-6 v2.0 expression beadchip ILMN_1653730 BC030015.1 human_transcriptome
Illumina human-6 v2.0 expression beadchip ILMN_1653730 NM_022120.1 human_transcriptome
Illumina human-6 v2.0 expression beadchip ILMN_1653730 AB050193.2 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1653730 BC072388.1 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1653730 AY013700.1 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1653730 BC030015.1 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1653730 NM_022120.1 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1653730 AB050193.2 human_transcriptome
Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_24641 - -
Illumina HumanRef-6 v2.0 expression beadchip ILMN_24641 - -
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1653730 BC072388.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1653730 AY013700.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1653730 BC030015.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1653730 NM_022120.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1653730 AB050193.2 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1653730 BC072388.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1653730 AY013700.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1653730 BC030015.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1653730 NM_022120.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1653730 AB050193.2 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1653730 BC072388.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1653730 AY013700.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1653730 BC030015.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1653730 NM_022120.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1653730 AB050193.2 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1653730 BC072388.1 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1653730 AY013700.1 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1653730 BC030015.1 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1653730 NM_022120.1 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1653730 AB050193.2 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1653730 BC072388.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1653730 AY013700.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1653730 BC030015.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1653730 NM_022120.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1653730 AB050193.2 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0