| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021473.hg.1 |
ENST00000161559.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021473.hg.1 |
ENST00000308072.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021473.hg.1 |
ENST00000351134.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021473.hg.1 |
ENST00000352591.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021473.hg.1 |
ENST00000358394.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021473.hg.1 |
ENST00000378065.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021473.hg.1 |
ENST00000403444.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021473.hg.1 |
ENST00000403461.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021473.hg.1 |
ENST00000446434.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021473.hg.1 |
ENST00000488639.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021473.hg.1 |
NM_001024912.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021473.hg.1 |
NM_001184813.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021473.hg.1 |
NM_001184815.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021473.hg.1 |
NM_001184816.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021473.hg.1 |
NM_001205344.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021473.hg.1 |
NM_001712.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021473.hg.1 |
TC19001575.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021468.hg.1 |
ENST00000161559.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021468.hg.1 |
ENST00000308072.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021468.hg.1 |
ENST00000351134.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021468.hg.1 |
ENST00000352591.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021468.hg.1 |
ENST00000358394.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021468.hg.1 |
ENST00000378065.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021468.hg.1 |
ENST00000403444.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021468.hg.1 |
ENST00000403461.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021468.hg.1 |
ENST00000446434.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021468.hg.1 |
ENST00000488639.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021468.hg.1 |
NM_001024912.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021468.hg.1 |
NM_001184813.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021468.hg.1 |
NM_001184815.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021468.hg.1 |
NM_001184816.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021468.hg.1 |
NM_001205344.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021468.hg.1 |
NM_001712.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021468.hg.1 |
TC19001575.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
ENST00000161559.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
ENST00000352591.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
ENST00000358394.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
ENST00000403444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
ENST00000403461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
ENST00000485605.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
ENST00000488639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
ENST00000599389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
NM_001024912.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
NM_001184813.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
NM_001184815.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
NM_001184816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
NM_001205344.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
NM_001712.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
TC1900010807.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
ENST00000161559.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
ENST00000352591.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
ENST00000358394.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
ENST00000403444.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
ENST00000403461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
ENST00000485605.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
ENST00000488639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
ENST00000599389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
NM_001024912.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
NM_001184813.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
NM_001184815.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
NM_001184816.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
NM_001205344.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189456.hg.1 |
NM_001712.4 |
Refseq |