This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00268409_m1
See other SSBP1 GE Assays ›
Gene Symbol:
SSBP1
Gene Name:
single stranded DNA binding protein 1
Gene Aliases:
Mt-SSB, SOSS-B1, SSBP, mtSSB
Chromosome Location:
Chr.7: 141738309 - 141750488 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00268409_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SSBP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001256510.1 NP_001243439.1 2 - 3 459 79
NM_001256511.1 NP_001243440.1 2 - 3 340 79
NM_001256512.1 NP_001243441.1 2 - 3 322 79
NM_001256513.1 NP_001243442.1 2 - 3 250 79
NM_003143.2 NP_003134.1 2 - 3 157 79
XM_005250048.4 XP_005250105.1 2 - 3 169 79
XM_005250049.4 XP_005250106.1 2 - 3 208 79
XM_005250050.4 XP_005250107.1 2 - 3 259 79
XM_005250051.4 XP_005250108.1 2 - 3 285 79
GenBank mRNA AF277319.1 - 2 - 3 103 79
AK294393.1 - 2 - 3 113 79
AK313033.1 - 2 - 3 170 79
BC000895.1 - 2 - 3 459 3464282 79
BC093054.1 - 2 - 3 119 30722138 79
BG030060.1 - 273 79
BQ959702.1 - 2 - 3 222 79
BU532904.1 - 2 - 3 160 79
DB464012.1 - 2 - 3 247 79
M94556.1 - 2 - 3 102 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700013469.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07000892.hg.1

Additional Information:

This assay is in the 5'UTR region of the AF277319.1,AK294393.1,AK313033.1,BC000895.1,BC093054.1,M94556.1,NM_001256510.1,NM_001256511.1,NM_001256512.1,NM_001256513.1,NM_003143.2,XM_005250048.4,XM_005250049.4,XM_005250050.4,XM_005250051.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SSBP1

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
mitochondrion organization
positive regulation of helicase activity
mitochondrion morphogenesis
chromatin binding
single-stranded DNA binding
protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000265304.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000265304.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000265304.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000265304.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000265304.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000265304.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000265304.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000461433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000461433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000461433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000461433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000463093.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000463093.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000463093.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000463093.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000463093.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000463093.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000463093.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000465167.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000465167.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000465167.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000465167.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000465167.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000465167.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000465167.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000467681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000467681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000467681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000467681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000467681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000467681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000467681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000467681.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000468267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000468267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000468267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000468267.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000469123.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000469123.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000469123.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000469123.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000469123.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000469123.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000469123.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000469123.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000473783.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000473783.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000473783.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000473783.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000481508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000481508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000481508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000481508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000481508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000481508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000481508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000481508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000481508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000481508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000484178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000484178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000484178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000484178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000484178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000484178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000484178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000484178.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000489378.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000489378.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000489378.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000489378.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000489378.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000489378.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000489378.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000496622.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000496622.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000496622.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000496622.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000498107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000498107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000498107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000498107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000498107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000498107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000498107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000612337.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000612337.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000612337.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000612337.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000612337.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000612337.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000612337.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000612337.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000612337.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 ENST00000612337.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_001256513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_003143.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_003143.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_003143.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_003143.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_003143.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_003143.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_003143.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NM_003143.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NR_046269.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NR_046269.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NR_046269.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NR_046269.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NR_046269.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NR_046269.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NR_046269.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NR_046269.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700013469.hg.1 NR_046269.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010189.hg.1 ENST00000441292.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200014364.hg.1 ENST00000415916.1 Ensembl
Affymetrix Human Clariom D Assay TC0200014364.hg.1 ENST00000415916.1 Ensembl
Affymetrix Human Clariom D Assay TC1000010189.hg.1 ENST00000441292.1 Ensembl
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 182993 - -
ABI Human Genome Survey Microarray Version 2.0 231126 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0