This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00269790_m1
See other GTF2F2 GE Assays ›
Gene Symbol:
GTF2F2
Gene Name:
general transcription factor IIF subunit 2
Gene Aliases:
BTF4, RAP30, TF2F2, TFIIF
Chromosome Location:
Chr.13: 45119830 - 45284105 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00269790_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GTF2F2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004128.2 NP_004119.1 1 - 2 226 71
XM_011535052.2 XP_011533354.1 1 - 2 258 71
XM_011535053.2 XP_011533355.1 1 - 2 258 71
XM_017020550.1 XP_016876039.1 1 - 2 258 71
GenBank mRNA AK291545.1 - 1 - 2 219 71
BC001771.1 - 1 - 2 187 3351446 71
BT019525.1 - 1 - 2 66 71
CB124466.1 - 1 - 2 226 71
DB252856.1 - 1 - 2 218 71
X16901.1 - 1 - 2 185 71
X59745.1 - 1 - 2 66 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300007052.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000171.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Human Transcription of mRNA 1700 Inventoried Human_Transcription_of_mRNA Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GTF2F2

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
7-methylguanosine mRNA capping
fibroblast growth factor receptor signaling pathway
gene expression
positive regulation of transcription elongation from RNA polymerase II promoter
snRNA transcription from RNA polymerase II promoter
positive regulation of RNA polymerase II transcriptional preinitiation complex assembly
positive regulation of viral transcription
positive regulation of transcription initiation from RNA polymerase II promoter
transcription factor activity, core RNA polymerase II binding
DNA binding
helicase activity
protein binding
ATP binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400008945.hg.1 ENST00000511083.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013836.hg.1 ENST00000445825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000461904.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000461904.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000461904.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000461904.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000461904.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000494087.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000494087.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000494087.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000494087.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 NM_004128.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 NM_004128.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 NM_004128.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 NM_004128.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 NM_004128.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 NM_004128.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 NM_004128.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 NM_004128.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 NM_004128.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 NM_004128.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 NM_004128.2 Refseq
Affymetrix Human Clariom D Assay TC0400008945.hg.1 ENST00000511083.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013836.hg.1 ENST00000445825.1 Ensembl
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 153454 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0