This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00271535_m1
See other MEF2A GE Assays ›
Gene Symbol:
MEF2A
Gene Name:
myocyte enhancer factor 2A
Gene Aliases:
ADCAD1, RSRFC4, RSRFC9, mef2
Chromosome Location:
Chr.15: 99565455 - 99716488 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00271535_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MEF2A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK294207.1 - 7 - 8 935 64
X68505.1 - 9 - 10 1302 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Human MAP Kinase Pathways 1700 Inventoried Human_Map_Kinase Probe spans exons

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 191872 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0