This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00287216_m1
See other MSANTD4 GE Assays ›
Gene Symbol:
MSANTD4
Gene Name:
Myb/SANT DNA binding domain containing 4 with coiled-coils
Gene Aliases:
KIAA1826
Chromosome Location:
Chr.11: 106007920 - 106022287 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00287216_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MSANTD4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_032424.2 NP_115800.1 1 - 2 1325 66
GenBank mRNA AB058729.1 - 1 - 2 1265 66
AK293050.1 - 1 - 2 121 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100012186.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11002254.hg.1

Additional Information:

This assay is in the 5'UTR region of the AB058729.1,AK293050.1,NM_032424.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MSANTD4

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11026960.hg.1 ENST00000301919.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11026960.hg.1 NM_032424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11026960.hg.1 TC11002254.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11026961.hg.1 ENST00000301919.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11026961.hg.1 NM_032424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11026961.hg.1 TC11002254.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000529805.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000529805.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000529805.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000530108.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000530108.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000530108.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000530108.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000530788.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000530788.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000530788.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000530788.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000530788.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000530788.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000534458.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000534458.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000534458.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000534458.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000617528.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000617528.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 ENST00000617528.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay PSR1100193750.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100193750.hg.1 ENST00000530108.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100193750.hg.1 ENST00000534458.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100193750.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay PSR1100193750.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100193750.hg.1 ENST00000530108.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100193750.hg.1 ENST00000534458.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100193750.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay PSR1100193750.hg.1 TC1100012186.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000529805.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000529805.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000529805.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000530108.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000530108.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000530108.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000530108.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000530788.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000530788.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000530788.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000530788.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000530788.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000530788.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000534458.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000534458.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000534458.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000534458.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000617528.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000617528.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 ENST00000617528.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay TC1100012186.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay PSR1100193746.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100193746.hg.1 ENST00000530108.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100193746.hg.1 ENST00000530788.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100193746.hg.1 ENST00000534458.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100193746.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay PSR1100193746.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100193746.hg.1 ENST00000530108.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100193746.hg.1 ENST00000530788.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100193746.hg.1 ENST00000534458.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100193746.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay PSR1100193746.hg.1 TC1100012186.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100193748.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100193748.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay PSR1100193748.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100193748.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay PSR1100193748.hg.1 TC1100012186.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100193749.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100193749.hg.1 ENST00000530108.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100193749.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay PSR1100193749.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100193749.hg.1 ENST00000530108.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100193749.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay PSR1100193749.hg.1 TC1100012186.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100193747.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100193747.hg.1 ENST00000530108.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100193747.hg.1 ENST00000530788.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100193747.hg.1 ENST00000534458.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100193747.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay PSR1100193747.hg.1 ENST00000301919.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100193747.hg.1 ENST00000530108.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100193747.hg.1 ENST00000530788.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100193747.hg.1 ENST00000534458.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100193747.hg.1 NM_032424.2 Refseq
Affymetrix Human Clariom D Assay PSR1100193747.hg.1 TC1100012186.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0