This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00290626_m1
See other TMEM155 GE Assays ›
Gene Symbol:
TMEM155
Gene Name:
transmembrane protein 155
Gene Aliases:
-
Chromosome Location:
Chr.4: 121758930 - 121765400 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00290626_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM155
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001317837.1 NP_001304766.1 2 - 3 327 62
NM_001317839.1 NP_001304768.1 2 - 3 202 62
NM_001317841.1 NP_001304770.1 2 - 3 483 62
NM_001317842.1 NP_001304771.1 2 - 3 327 62
NM_152399.3 NP_689612.3 2 - 3 483 62
XM_005262737.2 XP_005262794.1 2 - 3 291 62
XM_017007726.1 XP_016863215.1 2 - 3 200 62
GenBank mRNA AK055396.1 - 2 - 3 204 62
AK310583.1 - 2 - 3 202 62
BC047657.1 - 2 - 3 483 5264721 62
BX359599.2 - 2 - 3 355 62
DB116307.1 - 2 - 3 202 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400011746.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04001515.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK055396.1,BC047657.1,NM_001317837.1,NM_001317839.1,NM_001317841.1,NM_001317842.1,NM_152399.3,XM_005262737.2,XM_017007726.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TMEM155

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR04021517.hg.1 TC04001515.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04021517.hg.1 ENST00000337677.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04021517.hg.1 ENST00000394394.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04021517.hg.1 ENST00000394396.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04021517.hg.1 NM_152399.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000506551.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000506551.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000506551.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000506551.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000506551.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000514885.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000514885.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000514885.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000514885.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 ENST00000514885.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Clariom D Assay PSR0400180802.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Clariom D Assay PSR0400180802.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400180802.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400180802.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400180802.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400180802.hg.1 ENST00000506551.2 Ensembl
Affymetrix Human Clariom D Assay PSR0400180802.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400180802.hg.1 ENST00000514885.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400180802.hg.1 NM_152399.3 Refseq
Affymetrix Human Clariom D Assay PSR0400180802.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Clariom D Assay PSR0400180802.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400180802.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400180802.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400180802.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400180802.hg.1 ENST00000506551.2 Ensembl
Affymetrix Human Clariom D Assay PSR0400180802.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400180802.hg.1 ENST00000514885.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400180802.hg.1 NM_152399.3 Refseq
Affymetrix Human Clariom D Assay PSR0400180802.hg.1 TC0400011746.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000337677.9 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394394.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000394396.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000461198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000505198.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000506551.2 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000506551.2 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000506551.2 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000506551.2 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000506551.2 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000513254.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000514885.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000514885.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000514885.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000514885.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 ENST00000514885.5 Ensembl
Affymetrix Human Clariom D Assay TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Clariom D Assay TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Clariom D Assay TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Clariom D Assay TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Clariom D Assay TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Clariom D Assay TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Clariom D Assay TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Clariom D Assay TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Clariom D Assay TC0400011746.hg.1 NM_152399.3 Refseq
Affymetrix Human Clariom D Assay TC0400011746.hg.1 NM_152399.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0