This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00300599_m1
See other ZNF41 GE Assays ›
Gene Symbol:
ZNF41
Gene Name:
zinc finger protein 41
Gene Aliases:
MRX89
Chromosome Location:
Chr.X: 47444691 - 47483671 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00300599_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF41
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007130.3 NP_009061.1 1 - 2 488 139
XM_006724555.3 XP_006724618.1 1 - 2 469 139
XM_017029812.1 XP_016885301.1 1 - 2 469 139
GenBank mRNA AK290021.1 - 1 - 2 142 139
X60155.2 - 1 - 2 127 139

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00009569.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X000996.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK290021.1,NM_007130.3,X60155.2,XM_006724555.3,XM_017029812.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ZNF41

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X013214.hg.1 TC0X000996.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X013214.hg.1 ENST00000313116.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X013214.hg.1 ENST00000397050.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X013214.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X013213.hg.1 TC0X000996.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X013213.hg.1 ENST00000313116.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X013213.hg.1 ENST00000377065.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X013213.hg.1 ENST00000397050.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X013213.hg.1 NM_007130.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X013213.hg.1 NM_153380.3 Refseq
Affymetrix Human Clariom D Assay PSR0X00165220.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay PSR0X00165220.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay PSR0X00165220.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay PSR0X00165220.hg.1 NM_153380.3 Refseq
Affymetrix Human Clariom D Assay PSR0X00165220.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay PSR0X00165220.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay PSR0X00165220.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay PSR0X00165220.hg.1 NM_153380.3 Refseq
Affymetrix Human Clariom D Assay PSR0X00165220.hg.1 TC0X00009569.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0X00165221.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay PSR0X00165221.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay PSR0X00165221.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay PSR0X00165221.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay PSR0X00165221.hg.1 TC0X00009569.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000313116.11 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000377065.8 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000432977.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 ENST00000465311.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_007130.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_153380.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009569.hg.1 NM_153380.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0