This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00325982_m1
See other PNPLA8 GE Assays ›
Gene Symbol:
PNPLA8
Gene Name:
patatin like phospholipase domain containing 8
Gene Aliases:
IPLA2-2, IPLA2G, MMLA, PNPLA-gamma, iPLA2gamma
Chromosome Location:
Chr.7: 108470417 - 108528161 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00325982_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PNPLA8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001256008.2 NP_001242937.1 1 - 2 289 89
NM_001256009.2 NP_001242938.1 1 - 2 289 89
GenBank mRNA AB041261.1 - 1 - 2 115 89
AF263947.1 - 1 - 2 95 89
AL834147.1 - 1 - 2 42 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700013605.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07003301.hg.1

Additional Information:

This assay is in the 5'UTR region of the AB041261.1,AF263947.1,AL834147.1,NM_001256008.2,NM_001256009.2 transcripts.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Inventoried Probe spans exons

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

prostaglandin biosynthetic process
fatty acid metabolic process
cell death
arachidonic acid metabolic process
phosphatidylcholine catabolic process
phosphatidylcholine acyl-chain remodeling
phosphatidylethanolamine acyl-chain remodeling
linoleic acid metabolic process
phosphatidylethanolamine catabolic process
arachidonic acid secretion
lysophospholipase activity
phospholipase A2 activity
ATP binding
calcium-independent phospholipase A2 activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR07027400.hg.1 TC07003301.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07027400.hg.1 ENST00000257694.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07027400.hg.1 ENST00000388728.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07027400.hg.1 ENST00000422087.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07027400.hg.1 ENST00000426128.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07027400.hg.1 ENST00000436062.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07027400.hg.1 NM_001256007.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07027400.hg.1 NM_001256008.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07027400.hg.1 NM_001256009.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07027400.hg.1 NM_015723.4 Refseq
Affymetrix Human Clariom D Assay PSR0700201688.hg.1 ENST00000257694.12 Ensembl
Affymetrix Human Clariom D Assay PSR0700201688.hg.1 ENST00000415498.2 Ensembl
Affymetrix Human Clariom D Assay PSR0700201688.hg.1 ENST00000422087.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700201688.hg.1 ENST00000426128.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700201688.hg.1 ENST00000427008.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700201688.hg.1 ENST00000436062.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700201688.hg.1 NM_001256007.2 Refseq
Affymetrix Human Clariom D Assay PSR0700201688.hg.1 NM_001256008.2 Refseq
Affymetrix Human Clariom D Assay PSR0700201688.hg.1 NM_001256009.2 Refseq
Affymetrix Human Clariom D Assay PSR0700201688.hg.1 NM_015723.4 Refseq
Affymetrix Human Clariom D Assay PSR0700201688.hg.1 ENST00000257694.12 Ensembl
Affymetrix Human Clariom D Assay PSR0700201688.hg.1 ENST00000415498.2 Ensembl
Affymetrix Human Clariom D Assay PSR0700201688.hg.1 ENST00000422087.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700201688.hg.1 ENST00000426128.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700201688.hg.1 ENST00000427008.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700201688.hg.1 ENST00000436062.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700201688.hg.1 NM_001256007.2 Refseq
Affymetrix Human Clariom D Assay PSR0700201688.hg.1 NM_001256008.2 Refseq
Affymetrix Human Clariom D Assay PSR0700201688.hg.1 NM_001256009.2 Refseq
Affymetrix Human Clariom D Assay PSR0700201688.hg.1 NM_015723.4 Refseq
Affymetrix Human Clariom D Assay PSR0700201688.hg.1 TC0700013605.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0