This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00327177_m1
See other LRAT GE Assays ›
Gene Symbol:
LRAT
Gene Name:
lecithin retinol acyltransferase (phosphatidylcholine--retinol O-acyltransferase)
Gene Aliases:
LCA14
Chromosome Location:
Chr.4: 154740841 - 154753119 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00327177_m1
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LRAT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK056234.1 - 1 - 2 205 75
DQ266889.1 - 1 - 2 78 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Homo sapiens Inventoried Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000336356.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000336356.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000336356.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000336356.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000336356.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000336356.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000336356.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000336356.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000336356.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000499392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000499392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000499392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000499392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000499392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000499392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000500890.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000500890.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000500890.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000500890.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000502474.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000502474.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000502474.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000502474.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000502474.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000502474.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000502474.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000502525.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000502525.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000502525.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000502525.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000502525.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000507827.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000507827.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000507827.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000507827.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000507827.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000507827.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000507827.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000510733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000510733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000510733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000510733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000510733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000510733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000510733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000510733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000510733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000510919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000510919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000510919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000510919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000510919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 ENST00000510919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 NM_001301645.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 NM_001301645.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 NM_001301645.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 NM_001301645.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 NM_001301645.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 NM_001301645.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 NM_001301645.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 NM_001301645.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 NM_004744.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 NM_004744.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 NM_004744.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 NM_004744.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 NM_004744.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 NM_004744.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 NM_004744.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 NM_004744.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400009068.hg.1 NM_004744.4 Refseq
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000336356.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000336356.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000336356.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000336356.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000336356.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000336356.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000336356.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000336356.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000336356.3 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000499392.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000499392.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000499392.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000499392.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000499392.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000499392.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000500890.6 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000500890.6 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000500890.6 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000500890.6 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000502474.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000502474.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000502474.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000502474.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000502474.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000502474.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000502474.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000502525.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000502525.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000502525.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000502525.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000502525.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000507827.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000507827.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000507827.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000507827.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000507827.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000507827.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000507827.5 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000510733.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000510733.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000510733.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000510733.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000510733.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000510733.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000510733.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000510733.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000510733.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000510919.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000510919.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000510919.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000510919.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000510919.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 ENST00000510919.1 Ensembl
Affymetrix Human Clariom D Assay TC0400009068.hg.1 NM_001301645.1 Refseq
Affymetrix Human Clariom D Assay TC0400009068.hg.1 NM_001301645.1 Refseq
Affymetrix Human Clariom D Assay TC0400009068.hg.1 NM_001301645.1 Refseq
Affymetrix Human Clariom D Assay TC0400009068.hg.1 NM_001301645.1 Refseq
Affymetrix Human Clariom D Assay TC0400009068.hg.1 NM_001301645.1 Refseq
Affymetrix Human Clariom D Assay TC0400009068.hg.1 NM_001301645.1 Refseq
Affymetrix Human Clariom D Assay TC0400009068.hg.1 NM_001301645.1 Refseq
Affymetrix Human Clariom D Assay TC0400009068.hg.1 NM_001301645.1 Refseq
Affymetrix Human Clariom D Assay TC0400009068.hg.1 NM_004744.4 Refseq
Affymetrix Human Clariom D Assay TC0400009068.hg.1 NM_004744.4 Refseq
Affymetrix Human Clariom D Assay TC0400009068.hg.1 NM_004744.4 Refseq
Affymetrix Human Clariom D Assay TC0400009068.hg.1 NM_004744.4 Refseq
Affymetrix Human Clariom D Assay TC0400009068.hg.1 NM_004744.4 Refseq
Affymetrix Human Clariom D Assay TC0400009068.hg.1 NM_004744.4 Refseq
Affymetrix Human Clariom D Assay TC0400009068.hg.1 NM_004744.4 Refseq
Affymetrix Human Clariom D Assay TC0400009068.hg.1 NM_004744.4 Refseq
Affymetrix Human Clariom D Assay TC0400009068.hg.1 NM_004744.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0