This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00354988_m1
See other CDH2 GE Assays ›
Gene Symbol:
CDH2
Gene Name:
cadherin 2
Gene Aliases:
CD325, CDHN, CDw325, NCAD
Chromosome Location:
Chr.18: 27932878 - 28177446 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00354988_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDH2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001308176.1 NP_001295105.1 6 - 7 964 74
NM_001792.4 NP_001783.2 7 - 8 1444 74
XM_011525788.1 XP_011524090.1 7 - 8 1109 74
XM_017025514.1 XP_016881003.1 7 - 8 1415 74
GenBank mRNA BC036470.1 - 7 - 8 1136 5264944 74
M34064.1 - 4 - 5 543 74
S42303.1 - 7 - 8 1478 74
X57548.1 - 4 - 5 543 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1800008358.hg.1
This assay targets the same location as HTA transcript cluster(s) TC18000433.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CDH2

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
glial cell differentiation
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
cell migration
cerebral cortex development
regulation of myelination
adherens junction organization
regulation of Rho protein signal transduction
positive regulation of MAPK cascade
cell-cell adhesion mediated by cadherin
blood vessel morphogenesis
brain morphogenesis
homeostasis of number of cells
regulation of axonogenesis
striated muscle cell differentiation
positive regulation of muscle cell differentiation
protein heterooligomerization
radial glial cell differentiation
neuroepithelial cell differentiation
regulation of oligodendrocyte progenitor proliferation
establishment of protein localization to plasma membrane
negative regulation of canonical Wnt signaling pathway
neuroligin clustering involved in postsynaptic membrane assembly
neuronal stem cell population maintenance
regulation of postsynaptic density protein 95 clustering
positive regulation of synaptic vesicle clustering
calcium ion binding
protein binding
beta-catenin binding
protein kinase binding
protein phosphatase binding
protein complex binding
alpha-catenin binding
gamma-catenin binding
nitric-oxide synthase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000413878.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000413878.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000413878.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000413878.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000418492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000418492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000418492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000418492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000418492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000418492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000430882.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000430882.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000430882.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000430882.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 ENST00000430882.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18004393.hg.1 ENST00000269141.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18004393.hg.1 ENST00000399380.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18004393.hg.1 NM_001792.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18004393.hg.1 TC18000433.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1800156997.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay PSR1800156997.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Clariom D Assay PSR1800156997.hg.1 NM_001308176.1 Refseq
Affymetrix Human Clariom D Assay PSR1800156997.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay PSR1800156997.hg.1 TC1800008358.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1800156997.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay PSR1800156997.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Clariom D Assay PSR1800156997.hg.1 NM_001308176.1 Refseq
Affymetrix Human Clariom D Assay PSR1800156997.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000269141.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000399380.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000413878.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000413878.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000413878.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000413878.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000418492.5 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000418492.5 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000418492.5 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000418492.5 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000418492.5 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000418492.5 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000430882.5 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000430882.5 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000430882.5 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000430882.5 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 ENST00000430882.5 Ensembl
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001308176.1 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001792.4 Refseq
Affymetrix Human Clariom D Assay TC1800008358.hg.1 NM_001792.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline