This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00356080_g1
See other GSTM3 GE Assays ›
Gene Symbol:
GSTM3
Gene Name:
glutathione S-transferase mu 3
Gene Aliases:
GST5, GSTB, GSTM3-3, GTM3
Chromosome Location:
Chr.1: 109733932 - 109741038 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00356080_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GSTM3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000849.4 NP_000840.2 7 - 8 776 147
NR_024537.1 6 - 7 700 147
GenBank mRNA AK304300.1 - 5 - 6 428 147
AK313444.1 - 6 - 7 494 147
BC000088.2 - 6 - 7 481 3509784 147
BC008790.2 - 7 - 8 776 3630923 147
BT019945.1 - 6 - 7 466 147
CR542108.1 - 6 - 7 466 147
EF363096.1 - 6 - 7 483 147
J05459.1 - 6 - 7 483 147

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100015211.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01002962.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GSTM3

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
establishment of blood-nerve barrier
nitrobenzene metabolic process
xenobiotic catabolic process
response to estrogen
cellular detoxification of nitrogen compound
glutathione derivative biosynthetic process
glutathione transferase activity
enzyme binding
identical protein binding
protein homodimerization activity
glutathione binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01046054.hg.1 TC01002962.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01046054.hg.1 ENST00000256594.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01046054.hg.1 ENST00000361066.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01046054.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01046054.hg.1 ENST00000540225.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01046054.hg.1 NM_000849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01046054.hg.1 NR_024537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000476321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000476321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000476321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000476321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000476321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000476321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000476321.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000540225.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000540225.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000540225.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000540225.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000540225.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000540225.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000540225.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000540225.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 ENST00000540225.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Clariom D Assay PSR0100213383.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100213383.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100213383.hg.1 ENST00000476321.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100213383.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100213383.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100213383.hg.1 ENST00000540225.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100213383.hg.1 NM_000849.4 Refseq
Affymetrix Human Clariom D Assay PSR0100213383.hg.1 NR_024537.1 Refseq
Affymetrix Human Clariom D Assay PSR0100213383.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100213383.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100213383.hg.1 ENST00000476321.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100213383.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100213383.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100213383.hg.1 ENST00000540225.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100213383.hg.1 NM_000849.4 Refseq
Affymetrix Human Clariom D Assay PSR0100213383.hg.1 NR_024537.1 Refseq
Affymetrix Human Clariom D Assay PSR0100213383.hg.1 TC0100015211.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000256594.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000361066.6 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000476321.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000476321.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000476321.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000476321.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000476321.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000476321.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000476321.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000486823.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000488824.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000540225.2 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000540225.2 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000540225.2 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000540225.2 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000540225.2 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000540225.2 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000540225.2 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000540225.2 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 ENST00000540225.2 Ensembl
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NM_000849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NR_024537.1 Refseq
Affymetrix Human Clariom D Assay TC0100015211.hg.1 NR_024537.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0