This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00356285_g1
See other ADGRE5 GE Assays ›
Gene Symbol:
ADGRE5
Gene Name:
adhesion G protein-coupled receptor E5
Gene Aliases:
CD97, TM7LN1
Chromosome Location:
Chr.19: 14381144 - 14408725 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00356285_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRE5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025160.2 NP_001020331.1 15 - 16 2354 61
NM_001784.4 NP_001775.2 14 - 15 2222 61
NM_078481.3 NP_510966.1 16 - 17 2501 61
XM_011528451.2 XP_011526753.1 16 - 17 2453 61
XM_017027547.1 XP_016883036.1 17 - 18 2365 61
GenBank mRNA AK097264.1 - 14 - 15 1817 61
AK225655.1 - 15 - 16 2063 61
AK292159.1 - 14 - 15 2209 61
AK300343.1 - 15 - 16 1976 61
AK304553.1 - 12 - 13 1661 61
AK308368.1 - 14 - 15 2028 61
AK314697.1 - 16 - 17 2201 61
AX775807.1 - 1223 61
AX775809.1 - 1223 61
AX775811.1 - 14 - 15 1930 61
AX775813.1 - 16 - 17 2169 61
BC026690.2 - 14 - 15 1918 3847171 61
U76764.1 - 16 - 17 2169 61
X84700.1 - 14 - 15 1912 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900007173.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19000256.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ADGRE5

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

movement of cell or subcellular component
inflammatory response
immune response
cell adhesion
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
cell-cell signaling
transmembrane signaling receptor activity
G-protein coupled receptor activity
calcium ion binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR19003685.hg.1 ENST00000242786.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19003685.hg.1 ENST00000357355.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19003685.hg.1 ENST00000358600.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19003685.hg.1 ENST00000393059.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19003685.hg.1 NM_001025160.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19003685.hg.1 NM_001784.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19003685.hg.1 NM_078481.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19003685.hg.1 TC19000256.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1900151742.hg.1 ENST00000242786.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900151742.hg.1 ENST00000357355.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900151742.hg.1 ENST00000358600.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900151742.hg.1 NM_001025160.2 Refseq
Affymetrix Human Clariom D Assay PSR1900151742.hg.1 NM_001784.4 Refseq
Affymetrix Human Clariom D Assay PSR1900151742.hg.1 NM_078481.3 Refseq
Affymetrix Human Clariom D Assay PSR1900151742.hg.1 TC1900007173.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900151742.hg.1 ENST00000242786.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900151742.hg.1 ENST00000357355.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900151742.hg.1 ENST00000358600.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900151742.hg.1 NM_001025160.2 Refseq
Affymetrix Human Clariom D Assay PSR1900151742.hg.1 NM_001784.4 Refseq
Affymetrix Human Clariom D Assay PSR1900151742.hg.1 NM_078481.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0