This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00357218_g1
See other RPS19 GE Assays ›
Gene Symbol:
RPS19
Gene Name:
ribosomal protein S19
Gene Aliases:
DBA, DBA1, S19
Chromosome Location:
Chr.19: 41859918 - 41871416 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00357218_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPS19
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001022.3 NP_001013.1 1 - 2 378 134
NM_001321485.1 NP_001308414.1 1 - 2 378 134
XM_017027113.1 XP_016882602.1 1 - 2 373 134
GenBank mRNA BC000023.2 - 1 - 2 75 3504083 134
BC018616.1 - 1 - 2 378 4867036 134
CB124389.1 - 1 - 2 84 134

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900008164.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19000587.hg.1

Additional Information:

This assay is in the 5'UTR region of the BC000023.2,BC018616.1,NM_001022.3,NM_001321485.1,XM_017027113.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RPS19

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

ribosomal small subunit assembly
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
maturation of SSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
monocyte chemotaxis
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
nucleolus organization
Notch signaling pathway
response to extracellular stimulus
viral transcription
erythrocyte differentiation
maturation of SSU-rRNA
ribosomal small subunit biogenesis
protein tetramerization
positive regulation of cellular component movement
positive regulation of respiratory burst involved in inflammatory response
negative regulation of respiratory burst involved in inflammatory response
structural constituent of ribosome
protein binding
fibroblast growth factor binding
protein kinase binding
protein homodimerization activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR19008163.hg.1 ENST00000221975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19008163.hg.1 NM_001022.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19008163.hg.1 TC19000587.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000221975.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000221975.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000221975.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000221975.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000221975.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000221975.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000221975.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000221975.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000221975.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000593863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000593863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000593863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000593863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000593863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000593863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000593863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000593863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000593863.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598261.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598261.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598261.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598261.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598399.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598399.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598399.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598399.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598399.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598399.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598399.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000600467.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000600467.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000600467.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000600467.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000600467.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000601492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000601492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000601492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000601492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 ENST00000601492.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006832.hg.1 ENST00000415286.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006832.hg.1 ENST00000415286.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19008164.hg.1 ENST00000221975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19008164.hg.1 TC19000587.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR19008165.hg.1 ENST00000221975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19008165.hg.1 NM_001022.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19008165.hg.1 TC19000587.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1000010486.hg.1 ENST00000455650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008914.hg.1 ENST00000480487.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007375.hg.1 ENST00000477321.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007375.hg.1 ENST00000477321.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009470.hg.1 ENST00000457967.1 Ensembl
Affymetrix Human Clariom D Assay TC2000006832.hg.1 ENST00000415286.1 Ensembl
Affymetrix Human Clariom D Assay TC2000006832.hg.1 ENST00000415286.1 Ensembl
Affymetrix Human Clariom D Assay TC1000010486.hg.1 ENST00000455650.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900162172.hg.1 ENST00000221975.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900162172.hg.1 ENST00000598466.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162172.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162172.hg.1 ENST00000600467.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162172.hg.1 ENST00000601492.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162172.hg.1 NM_001022.3 Refseq
Affymetrix Human Clariom D Assay PSR1900162172.hg.1 TC1900008164.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900162172.hg.1 ENST00000221975.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900162172.hg.1 ENST00000598466.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162172.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162172.hg.1 ENST00000600467.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162172.hg.1 ENST00000601492.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162172.hg.1 NM_001022.3 Refseq
Affymetrix Human Clariom D Assay PSR1900162169.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162169.hg.1 NM_001022.3 Refseq
Affymetrix Human Clariom D Assay PSR1900162169.hg.1 TC1900008164.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900162169.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162169.hg.1 NM_001022.3 Refseq
Affymetrix Human Clariom D Assay TC1400008914.hg.1 ENST00000480487.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000221975.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000221975.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000221975.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000221975.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000221975.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000221975.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000221975.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000221975.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000221975.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000593863.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000593863.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000593863.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000593863.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000593863.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000593863.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000593863.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000593863.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000593863.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598261.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598261.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598261.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598261.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598399.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598399.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598399.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598399.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598399.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598399.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598399.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598466.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598466.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598466.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598466.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598466.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598466.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000600467.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000600467.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000600467.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000600467.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000600467.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000601492.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000601492.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000601492.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000601492.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 ENST00000601492.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Clariom D Assay TC1900008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Clariom D Assay TC1900008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Clariom D Assay TC1900008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Clariom D Assay TC1900008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Clariom D Assay TC1900008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Clariom D Assay TC1900008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Clariom D Assay TC1900008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Clariom D Assay TC1900008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Clariom D Assay TC1900008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Clariom D Assay TC1900008164.hg.1 NM_001022.3 Refseq
Affymetrix Human Clariom D Assay PSR1900162170.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162170.hg.1 NM_001022.3 Refseq
Affymetrix Human Clariom D Assay PSR1900162170.hg.1 TC1900008164.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900162170.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162170.hg.1 NM_001022.3 Refseq
Affymetrix Human Clariom D Assay PSR1900162178.hg.1 ENST00000221975.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900162178.hg.1 ENST00000593863.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162178.hg.1 ENST00000598261.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900162178.hg.1 ENST00000598399.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900162178.hg.1 ENST00000598466.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162178.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162178.hg.1 ENST00000600467.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162178.hg.1 ENST00000601492.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162178.hg.1 NM_001022.3 Refseq
Affymetrix Human Clariom D Assay PSR1900162178.hg.1 TC1900008164.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900162178.hg.1 ENST00000221975.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900162178.hg.1 ENST00000593863.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162178.hg.1 ENST00000598261.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900162178.hg.1 ENST00000598399.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900162178.hg.1 ENST00000598466.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162178.hg.1 ENST00000598742.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162178.hg.1 ENST00000600467.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162178.hg.1 ENST00000601492.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900162178.hg.1 NM_001022.3 Refseq
Affymetrix Human Clariom D Assay TC0100009470.hg.1 ENST00000457967.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007375.hg.1 ENST00000477321.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007375.hg.1 ENST00000477321.1 Ensembl
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 184007 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rp79v:80/100.66.77.211:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline