This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00357983_g1
See other MPG GE Assays ›
Gene Symbol:
MPG
Gene Name:
N-methylpurine DNA glycosylase
Gene Aliases:
AAG, ADPG, APNG, CRA36.1, MDG, Mid1, PIG11, PIG16, anpg
Chromosome Location:
Chr.16: 77019 - 85851 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00357983_g1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MPG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002434.3 NP_002425.2 2 - 3 353 68
GenBank mRNA AL520052.3 - 2 - 3 264 68
M74905.1 - 1 - 2 181 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600006439.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000004.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Human Estrogen Panel Inventoried Human_Estrogen Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MPG

Panther Classification:

Molecular Function -

DNA metabolism protein DNA glycosylase nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

base-excision repair
DNA dealkylation involved in DNA repair
depurination
damaged DNA binding
alkylbase DNA N-glycosylase activity
protein binding
DNA-3-methyladenine glycosylase activity
DNA N-glycosylase activity
DNA-7-methylguanine glycosylase activity
DNA-7-methyladenine glycosylase activity
DNA-3-methylguanine glycosylase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16000048.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16000048.hg.1 ENST00000356432.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16000048.hg.1 ENST00000397817.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16000048.hg.1 ENST00000475280.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16000048.hg.1 NM_001015052.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16000048.hg.1 NM_001015054.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16000048.hg.1 NM_002434.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16000048.hg.1 TC16000004.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR16000047.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16000047.hg.1 NM_002434.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16000047.hg.1 TC16000004.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR16000046.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16000046.hg.1 ENST00000356432.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16000046.hg.1 ENST00000475280.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16000046.hg.1 NM_001015052.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16000046.hg.1 NM_002434.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16000046.hg.1 TC16000004.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000356432.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000356432.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000356432.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000356432.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000356432.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000356432.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000356432.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000356432.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000397817.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000397817.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000397817.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000397817.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000397817.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000397817.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000436333.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000436333.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000436333.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000436333.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000475280.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000475280.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 ENST00000475280.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 NM_001015052.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 NM_001015052.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 NM_001015052.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 NM_001015052.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 NM_001015052.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 NM_001015052.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 NM_001015052.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 NM_001015054.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 NM_001015054.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 NM_001015054.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 NM_001015054.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 NM_001015054.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 NM_001015054.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 NM_002434.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 NM_002434.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 NM_002434.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 NM_002434.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 NM_002434.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 NM_002434.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 NM_002434.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 NM_002434.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006439.hg.1 NM_002434.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16000043.hg.1 ENST00000356432.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16000043.hg.1 NM_001015052.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16000043.hg.1 NM_002434.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16000043.hg.1 TC16000004.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000356432.7 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000356432.7 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000356432.7 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000356432.7 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000356432.7 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000356432.7 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000356432.7 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000356432.7 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000397817.5 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000397817.5 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000397817.5 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000397817.5 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000397817.5 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000397817.5 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000436333.5 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000436333.5 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000436333.5 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000436333.5 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000475280.1 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000475280.1 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 ENST00000475280.1 Ensembl
Affymetrix Human Clariom D Assay TC1600006439.hg.1 NM_001015052.2 Refseq
Affymetrix Human Clariom D Assay TC1600006439.hg.1 NM_001015052.2 Refseq
Affymetrix Human Clariom D Assay TC1600006439.hg.1 NM_001015052.2 Refseq
Affymetrix Human Clariom D Assay TC1600006439.hg.1 NM_001015052.2 Refseq
Affymetrix Human Clariom D Assay TC1600006439.hg.1 NM_001015052.2 Refseq
Affymetrix Human Clariom D Assay TC1600006439.hg.1 NM_001015052.2 Refseq
Affymetrix Human Clariom D Assay TC1600006439.hg.1 NM_001015052.2 Refseq
Affymetrix Human Clariom D Assay TC1600006439.hg.1 NM_001015054.2 Refseq
Affymetrix Human Clariom D Assay TC1600006439.hg.1 NM_001015054.2 Refseq
Affymetrix Human Clariom D Assay TC1600006439.hg.1 NM_001015054.2 Refseq
Affymetrix Human Clariom D Assay TC1600006439.hg.1 NM_001015054.2 Refseq
Affymetrix Human Clariom D Assay TC1600006439.hg.1 NM_001015054.2 Refseq
Affymetrix Human Clariom D Assay TC1600006439.hg.1 NM_001015054.2 Refseq
Affymetrix Human Clariom D Assay TC1600006439.hg.1 NM_002434.3 Refseq
Affymetrix Human Clariom D Assay TC1600006439.hg.1 NM_002434.3 Refseq
Affymetrix Human Clariom D Assay TC1600006439.hg.1 NM_002434.3 Refseq
Affymetrix Human Clariom D Assay TC1600006439.hg.1 NM_002434.3 Refseq
Affymetrix Human Clariom D Assay TC1600006439.hg.1 NM_002434.3 Refseq
Affymetrix Human Clariom D Assay TC1600006439.hg.1 NM_002434.3 Refseq
Affymetrix Human Clariom D Assay TC1600006439.hg.1 NM_002434.3 Refseq
Affymetrix Human Clariom D Assay TC1600006439.hg.1 NM_002434.3 Refseq
Affymetrix Human Clariom D Assay TC1600006439.hg.1 NM_002434.3 Refseq
Affymetrix Human Clariom D Assay PSR1600144109.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Clariom D Assay PSR1600144109.hg.1 NM_002434.3 Refseq
Affymetrix Human Clariom D Assay PSR1600144109.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Clariom D Assay PSR1600144109.hg.1 NM_002434.3 Refseq
Affymetrix Human Clariom D Assay PSR1600144109.hg.1 TC1600006439.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600144110.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Clariom D Assay PSR1600144110.hg.1 NM_002434.3 Refseq
Affymetrix Human Clariom D Assay PSR1600144110.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Clariom D Assay PSR1600144110.hg.1 NM_002434.3 Refseq
Affymetrix Human Clariom D Assay PSR1600144110.hg.1 TC1600006439.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600144104.hg.1 ENST00000356432.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600144104.hg.1 NM_001015052.2 Refseq
Affymetrix Human Clariom D Assay PSR1600144104.hg.1 NM_002434.3 Refseq
Affymetrix Human Clariom D Assay PSR1600144104.hg.1 ENST00000356432.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600144104.hg.1 NM_001015052.2 Refseq
Affymetrix Human Clariom D Assay PSR1600144104.hg.1 NM_002434.3 Refseq
Affymetrix Human Clariom D Assay PSR1600144104.hg.1 TC1600006439.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600144107.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Clariom D Assay PSR1600144107.hg.1 ENST00000356432.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600144107.hg.1 ENST00000475280.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600144107.hg.1 NM_001015052.2 Refseq
Affymetrix Human Clariom D Assay PSR1600144107.hg.1 NM_002434.3 Refseq
Affymetrix Human Clariom D Assay PSR1600144107.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Clariom D Assay PSR1600144107.hg.1 ENST00000356432.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600144107.hg.1 ENST00000475280.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600144107.hg.1 NM_001015052.2 Refseq
Affymetrix Human Clariom D Assay PSR1600144107.hg.1 NM_002434.3 Refseq
Affymetrix Human Clariom D Assay PSR1600144107.hg.1 TC1600006439.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600144112.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Clariom D Assay PSR1600144112.hg.1 ENST00000356432.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600144112.hg.1 ENST00000397817.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600144112.hg.1 ENST00000436333.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600144112.hg.1 ENST00000475280.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600144112.hg.1 NM_001015052.2 Refseq
Affymetrix Human Clariom D Assay PSR1600144112.hg.1 NM_001015054.2 Refseq
Affymetrix Human Clariom D Assay PSR1600144112.hg.1 NM_002434.3 Refseq
Affymetrix Human Clariom D Assay PSR1600144112.hg.1 ENST00000219431.4 Ensembl
Affymetrix Human Clariom D Assay PSR1600144112.hg.1 ENST00000356432.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600144112.hg.1 ENST00000397817.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600144112.hg.1 ENST00000436333.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600144112.hg.1 ENST00000475280.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600144112.hg.1 NM_001015052.2 Refseq
Affymetrix Human Clariom D Assay PSR1600144112.hg.1 NM_001015054.2 Refseq
Affymetrix Human Clariom D Assay PSR1600144112.hg.1 NM_002434.3 Refseq
Affymetrix Human Clariom D Assay PSR1600144112.hg.1 TC1600006439.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0