This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00364160_g1
See other SLC7A3 GE Assays ›
Gene Symbol:
SLC7A3
Gene Name:
solute carrier family 7 member 3
Gene Aliases:
ATRC3, CAT-3, CAT3
Chromosome Location:
Chr.X: 70925579 - 70931125 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00364160_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC7A3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001048164.2 NP_001041629.1 4 - 5 859 70
NM_032803.5 NP_116192.4 4 - 5 883 70
XM_017029912.1 XP_016885401.1 3 - 4 1374 70
GenBank mRNA AF320612.1 - 4 - 5 834 70
AK027447.1 - 2 - 3 205 70
AK074865.1 - 4 - 5 854 70
AK075014.1 - 4 - 5 859 70
BC033816.1 - 4 - 5 838 5206252 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00009972.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X001117.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC7A3

Panther Classification:

Molecular Function -

amino acid transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transport
arginine transport
lysine transport
ornithine transport
regulation of TOR signaling
L-ornithine transmembrane transport
L-lysine transmembrane transport
arginine transmembrane transport
basic amino acid transmembrane transport
L-ornithine transmembrane transporter activity
amino acid transmembrane transporter activity
arginine transmembrane transporter activity
L-lysine transmembrane transporter activity
antiporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR0X015206.hg.1 TC0X001117.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X015206.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X015206.hg.1 ENST00000374299.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X015206.hg.1 NM_001048164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X015206.hg.1 NM_032803.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_001048164.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Clariom D Assay TC0X00009972.hg.1 NM_032803.5 Refseq
Affymetrix Human Clariom D Assay PSR0X00168095.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Clariom D Assay PSR0X00168095.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00168095.hg.1 NM_001048164.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00168095.hg.1 NM_032803.5 Refseq
Affymetrix Human Clariom D Assay PSR0X00168095.hg.1 ENST00000298085.4 Ensembl
Affymetrix Human Clariom D Assay PSR0X00168095.hg.1 ENST00000374299.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00168095.hg.1 NM_001048164.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00168095.hg.1 NM_032803.5 Refseq
Affymetrix Human Clariom D Assay PSR0X00168095.hg.1 TC0X00009972.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0