This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00365485_m1
See other VCAM1 GE Assays ›
Gene Symbol:
VCAM1
Gene Name:
vascular cell adhesion molecule 1
Gene Aliases:
CD106, INCAM-100
Chromosome Location:
Chr.1: 100719640 - 100739045 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00365485_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VCAM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001078.3 NP_001069.1 7 - 8 2018 84
NM_001199834.1 NP_001186763.1 7 - 8 1832 84
NM_080682.2 NP_542413.1 6 - 7 1742 84
GenBank mRNA AK223266.1 - 7 - 8 1917 84
AK291732.1 - 7 - 8 1916 84
AK296692.1 - 7 - 8 1730 84
BC017276.2 - 7 - 8 1898 4845424 84
BC068490.1 - 7 - 8 1916 30339170 84
BC085003.1 - 7 - 8 1898 6375943 84
M30257.1 - 6 - 7 1627 84
M60335.1 - 7 - 8 1896 84
X53051.1 - 7 - 8 1797 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100009221.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000904.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene VCAM1

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
acute inflammatory response
chronic inflammatory response
cell adhesion
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
leukocyte cell-cell adhesion
cell-matrix adhesion
aging
response to nutrient
amine metabolic process
response to zinc ion
response to ionizing radiation
viral process
membrane to membrane docking
B cell differentiation
extracellular matrix organization
response to lipopolysaccharide
response to nicotine
calcium-mediated signaling using intracellular calcium source
cellular response to vascular endothelial growth factor stimulus
positive regulation of T cell proliferation
response to ethanol
regulation of immune response
leukocyte tethering or rolling
oxidation-reduction process
cell chemotaxis
interferon-gamma-mediated signaling pathway
cellular response to tumor necrosis factor
integrin binding
primary amine oxidase activity
cell adhesion molecule binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01014257.hg.1 TC01000904.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01014257.hg.1 ENST00000294728.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01014257.hg.1 ENST00000347652.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01014257.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01014257.hg.1 ENST00000370119.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01014257.hg.1 NM_001078.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01014257.hg.1 NM_001199834.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01014257.hg.1 NM_080682.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000603679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000603679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000603679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 ENST00000603679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000603679.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000603679.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000603679.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 ENST00000603679.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001078.3 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_001199834.1 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Clariom D Assay TC0100009221.hg.1 NM_080682.2 Refseq
Affymetrix Human Clariom D Assay PSR0100165692.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100165692.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100165692.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100165692.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100165692.hg.1 NM_001078.3 Refseq
Affymetrix Human Clariom D Assay PSR0100165692.hg.1 NM_001199834.1 Refseq
Affymetrix Human Clariom D Assay PSR0100165692.hg.1 NM_080682.2 Refseq
Affymetrix Human Clariom D Assay PSR0100165692.hg.1 ENST00000294728.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100165692.hg.1 ENST00000347652.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100165692.hg.1 ENST00000370115.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100165692.hg.1 ENST00000370119.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100165692.hg.1 NM_001078.3 Refseq
Affymetrix Human Clariom D Assay PSR0100165692.hg.1 NM_001199834.1 Refseq
Affymetrix Human Clariom D Assay PSR0100165692.hg.1 NM_080682.2 Refseq
Affymetrix Human Clariom D Assay PSR0100165692.hg.1 TC0100009221.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0