This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00365600_s1
See other SOCS4 GE Assays ›
Gene Symbol:
SOCS4
Gene Name:
suppressor of cytokine signaling 4
Gene Aliases:
SOCS7
Chromosome Location:
Chr.14: 55027126 - 55049489 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00365600_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 330.60
330.6
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SOCS4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080867.2 NP_543143.1 2 2881 129
NM_199421.1 NP_955453.1 3 3010 129
XM_011536425.1 XP_011534727.1 3 3064 129
XM_011536426.1 XP_011534728.1 3 2979 129
GenBank mRNA AF424815.1 - 1 2661 129
AK127299.1 - 1 1759 129
BC060790.1 - 3 2888 30338646 129
BX648734.1 - 2 2667 129
CR933659.1 - 2 2775 129

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1400007221.hg.1
This assay targets the same location as HTA transcript cluster(s) TC14000321.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SOCS4

Panther Classification:

Molecular Function -

kinase modulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of epidermal growth factor-activated receptor activity
protein ubiquitination
cytokine-mediated signaling pathway
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
intracellular signal transduction
regulation of growth
negative regulation of JAK-STAT cascade
protein kinase inhibitor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000553735.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000553735.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14003425.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14003425.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14003425.hg.1 NM_080867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14003425.hg.1 NM_199421.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14003425.hg.1 TC14000321.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000553735.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000553735.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Clariom D Assay PSR1400150053.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Clariom D Assay PSR1400150053.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Clariom D Assay PSR1400150053.hg.1 NM_080867.2 Refseq
Affymetrix Human Clariom D Assay PSR1400150053.hg.1 NM_199421.1 Refseq
Affymetrix Human Clariom D Assay PSR1400150053.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Clariom D Assay PSR1400150053.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Clariom D Assay PSR1400150053.hg.1 NM_080867.2 Refseq
Affymetrix Human Clariom D Assay PSR1400150053.hg.1 NM_199421.1 Refseq
Affymetrix Human Clariom D Assay PSR1400150053.hg.1 TC1400007221.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-5xwv5:80/100.66.76.242:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0