This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00368972_m1
See other METTL26 GE Assays ›
Gene Symbol:
METTL26
Gene Name:
methyltransferase like 26
Gene Aliases:
C16orf13, JFP2
Chromosome Location:
Chr.16: 634427 - 636366 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00368972_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: METTL26
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001040161.2 NP_001035251.1 1 - 2 276 101
GenBank mRNA BC007207.1 - 1 - 2 258 4054799 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600008963.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000731.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16010624.hg.1 ENST00000301686.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16010624.hg.1 ENST00000338401.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16010624.hg.1 ENST00000397664.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16010624.hg.1 ENST00000397665.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16010624.hg.1 ENST00000397666.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16010624.hg.1 ENST00000456420.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16010624.hg.1 NM_001040160.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16010624.hg.1 NM_001040161.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16010624.hg.1 NM_001040162.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16010624.hg.1 NM_001040165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16010624.hg.1 NM_032366.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16010624.hg.1 TC16000731.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR16010635.hg.1 ENST00000301686.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16010635.hg.1 ENST00000338401.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16010635.hg.1 ENST00000397664.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16010635.hg.1 ENST00000397665.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16010635.hg.1 ENST00000397666.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16010635.hg.1 NM_001040160.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16010635.hg.1 NM_001040161.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16010635.hg.1 NM_001040162.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16010635.hg.1 NM_001040165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16010635.hg.1 NM_032366.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16010635.hg.1 TC16000731.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR16010633.hg.1 ENST00000301686.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16010633.hg.1 ENST00000338401.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16010633.hg.1 ENST00000397664.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16010633.hg.1 ENST00000397665.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16010633.hg.1 ENST00000397666.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16010633.hg.1 NM_001040160.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16010633.hg.1 NM_001040161.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16010633.hg.1 NM_001040162.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16010633.hg.1 NM_001040165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16010633.hg.1 NM_032366.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16010633.hg.1 TC16000731.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000301686.12 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000338401.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000397664.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000397665.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000397666.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000448973.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000456420.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000564039.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000565163.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000565799.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000568077.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000568773.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000568830.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000614890.4 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 NM_001040160.2 Refseq
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 NM_001040161.2 Refseq
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 NM_001040162.2 Refseq
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 NM_001040165.2 Refseq
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 NM_001288710.1 Refseq
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 NM_032366.4 Refseq
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 NR_109976.1 Refseq
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 NR_109977.1 Refseq
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 NR_109978.1 Refseq
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 NR_109979.1 Refseq
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000301686.12 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000338401.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000397664.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000397665.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000397666.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000448973.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000456420.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000564039.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000565163.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000565799.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000568077.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000568773.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000568830.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 ENST00000614890.4 Ensembl
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 NM_001040160.2 Refseq
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 NM_001040161.2 Refseq
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 NM_001040162.2 Refseq
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 NM_001040165.2 Refseq
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 NM_001288710.1 Refseq
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 NM_032366.4 Refseq
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 NR_109976.1 Refseq
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 NR_109977.1 Refseq
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 NR_109978.1 Refseq
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 NR_109979.1 Refseq
Affymetrix Human Clariom D Assay PSR1600167627.hg.1 TC1600008963.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 121186 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0