This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00369138_m1
See other ZNF160 GE Assays ›
Gene Symbol:
ZNF160
Gene Name:
zinc finger protein 160
Gene Aliases:
F11, HKr18, HZF5, KR18
Chromosome Location:
Chr.19: 53064882 - 53103434 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00369138_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF160
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001102603.1 NP_001096073.1 1 - 2 168 73
NM_001322125.1 NP_001309054.1 1 - 2 168 73
NM_001322126.1 NP_001309055.1 1 - 2 168 73
NM_001322128.1 NP_001309057.1 1 - 2 168 73
NM_001322129.1 NP_001309058.1 1 - 2 168 73
NM_001322130.1 NP_001309059.1 1 - 2 168 73
NM_001322131.1 NP_001309060.1 1 - 2 168 73
NM_001322132.1 NP_001309061.1 1 - 2 168 73
NM_001322133.1 NP_001309062.1 1 - 2 168 73
NM_001322134.1 NP_001309063.1 1 - 2 168 73
NM_001322135.1 NP_001309064.1 1 - 2 168 73
NM_001322137.1 NP_001309066.1 1 - 2 168 73
NM_001322138.1 NP_001309067.1 1 - 2 168 73
NM_001322139.1 NP_001309068.1 1 - 2 168 73
NM_033288.3 NP_150630.2 1 - 2 168 73
NM_198893.2 NP_942596.1 1 - 2 168 73
GenBank mRNA AF277623.1 - 1 - 2 176 73
BC000807.1 - 1 - 2 137 73
BC094880.1 - 1 - 2 85 3066532 73
BU430880.1 - 1 - 2 92 73
DA230397.1 - 1 - 2 230 73
DB078640.1 - 1 - 2 168 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900011327.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19001811.hg.1

Additional Information:

This assay is in the 5'UTR region of the AF277623.1,BC000807.1,BC094880.1,NM_001102603.1,NM_001322125.1,NM_001322126.1,NM_001322128.1,NM_001322129.1,NM_001322130.1,NM_001322131.1,NM_001322132.1,NM_001322133.1,NM_001322134.1,NM_001322135.1,NM_001322137.1,NM_001322138.1,NM_001322139.1,NM_033288.3,NM_198893.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ZNF160

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
hemopoiesis
DNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000355147.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000355147.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000355147.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000355147.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000355147.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000355147.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000355147.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000355147.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000355147.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000355147.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000418871.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000418871.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000418871.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000418871.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000418871.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000418871.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000418871.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000418871.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000418871.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000418871.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000418871.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000418871.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000418871.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000429604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000429604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000429604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000429604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000429604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000429604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000429604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000429604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000429604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000429604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000429604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000429604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000429604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000429604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000429604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000594760.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000594760.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000594760.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000594760.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000594760.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000594760.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000594760.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000594760.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000596388.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000596388.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000596388.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000596966.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000596966.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000596966.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000596966.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000596966.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000596966.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000596966.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000596966.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000597112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000597112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000597112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000597112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000597112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000597112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000597112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000597112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000597112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000597112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000597112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000597112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000597112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000597112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000597112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599056.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599056.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599056.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599056.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599056.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599056.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599056.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599056.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599056.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599056.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599056.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599056.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599056.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599247.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599247.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599247.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599247.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599247.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599247.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599247.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599247.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599247.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599637.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599637.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599637.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599637.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599637.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599637.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599637.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599637.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599729.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599729.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599729.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599729.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599980.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599980.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599980.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599980.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599980.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000599980.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000600924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000600924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000600924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000600924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000600924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000600924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000600924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000600924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000601421.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000601421.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000601421.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000601421.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000601421.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000601421.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000601421.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000601421.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000601421.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000601421.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000601982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000601982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000601982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000601982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000601982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000601982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000602095.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000602095.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000602095.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000602095.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000602095.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000602095.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000602095.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 ENST00000602095.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_001102603.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_001102603.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_001102603.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_001102603.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_001102603.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_001102603.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_001102603.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_001102603.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_001102603.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_001102603.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_001102603.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_001102603.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_001102603.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_001102603.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_001102603.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_033288.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_033288.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_033288.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_033288.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_033288.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_033288.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_033288.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_033288.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_033288.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_033288.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_033288.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_033288.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_033288.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_198893.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_198893.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_198893.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_198893.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_198893.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_198893.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_198893.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_198893.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_198893.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_198893.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_198893.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_198893.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_198893.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011327.hg.1 NM_198893.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0