This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00369695_m1
See other MOGAT1 GE Assays ›
Gene Symbol:
MOGAT1
Gene Name:
monoacylglycerol O-acyltransferase 1
Gene Aliases:
DGAT2L, DGAT2L1, MGAT1
Chromosome Location:
Chr.2: 222671738 - 222709930 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00369695_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MOGAT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_058165.2 NP_477513.2 3 - 4 529 75
GenBank mRNA AF384163.1 - 3 - 4 526 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200010902.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001349.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MOGAT1

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

glycerol metabolic process
diacylglycerol biosynthetic process
triglyceride biosynthetic process
2-acylglycerol O-acyltransferase activity
diacylglycerol O-acyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02021039.hg.1 TC02001349.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02021039.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02021039.hg.1 NM_058165.2 Refseq
Affymetrix Human Clariom D Assay PSR0200178156.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Clariom D Assay PSR0200178156.hg.1 NM_058165.2 Refseq
Affymetrix Human Clariom D Assay PSR0200178156.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Clariom D Assay PSR0200178156.hg.1 NM_058165.2 Refseq
Affymetrix Human Clariom D Assay PSR0200178156.hg.1 TC0200010902.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Clariom D Assay TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Clariom D Assay TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Clariom D Assay TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Clariom D Assay TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Clariom D Assay TC0200010902.hg.1 ENST00000446656.3 Ensembl
Affymetrix Human Clariom D Assay TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Clariom D Assay TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Clariom D Assay TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Clariom D Assay TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Clariom D Assay TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Clariom D Assay TC0200010902.hg.1 NM_058165.2 Refseq
Affymetrix Human Clariom D Assay TC0200010902.hg.1 NM_058165.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0