This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00371445_g1
See other DCHS1 GE Assays ›
Gene Symbol:
DCHS1
Gene Name:
dachsous cadherin-related 1
Gene Aliases:
CDH19, CDH25, CDHR6, FIB1, MVP2, PCDH16, VMLDS1
Chromosome Location:
Chr.11: 6621324 - 6655849 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00371445_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DCHS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003737.3 NP_003728.1 11 - 12 5441 84
GenBank mRNA AB053446.1 - 11 - 12 5444 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100009982.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11001363.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DCHS1

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

branching involved in ureteric bud morphogenesis
mitral valve formation
cell migration involved in endocardial cushion formation
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
neural tube development
neurogenesis
hippo signaling
post-anal tail morphogenesis
ossification involved in bone maturation
digestive tract development
cochlea development
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11017174.hg.1 ENST00000299441.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11017174.hg.1 NM_003737.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11017174.hg.1 TC11001363.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay TC1100009982.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay PSR1100175462.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100175462.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay PSR1100175462.hg.1 ENST00000299441.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100175462.hg.1 NM_003737.3 Refseq
Affymetrix Human Clariom D Assay PSR1100175462.hg.1 TC1100009982.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0