This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00372938_m1
See other BMF GE Assays ›
Gene Symbol:
BMF
Gene Name:
Bcl2 modifying factor
Gene Aliases:
-
Chromosome Location:
Chr.15: 40087890 - 40108884 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00372938_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BMF
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003940.1 NP_001003940.1 4 - 5 689 59
NM_033503.3 NP_277038.1 3 - 4 540 59
GenBank mRNA AK024472.1 - 3 - 4 550 59
AY029254.1 - 2 - 3 454 59
BC060783.1 - 4 - 5 687 59
BC069328.1 - 3 - 4 578 7262161 59
BC069505.1 - 3 - 4 524 7262173 59
BC070043.1 - 4 - 5 687 30344794 59
BC104983.1 - 3 - 4 475 8143986 59
BC104985.1 - 3 - 4 475 8143988 59

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1500009082.hg.1
This assay targets the same location as HTA transcript cluster(s) TC15001230.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene BMF

Gene Ontology Categories:

Function(s) Process(es)

protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
positive regulation of protein homooligomerization
positive regulation of apoptotic process
anoikis
positive regulation of release of cytochrome c from mitochondria
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
positive regulation of intrinsic apoptotic signaling pathway
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000354670.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000354670.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000354670.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000354670.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000354670.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000354670.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000354670.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000354670.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000354670.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000354670.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000354670.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000354670.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000397573.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000397573.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000397573.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000397573.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000397573.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000397573.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000397573.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000397573.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000397573.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000397573.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000431415.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000431415.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000431415.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000431415.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000431415.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000557870.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000557870.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000557870.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000557870.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000557870.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000557870.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000558057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000558057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000558057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000558057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000558774.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000558774.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000558774.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000558774.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000558774.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000558774.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000558774.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000559701.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000559701.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000559701.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000559701.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000559701.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000559701.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000559701.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000560430.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000560430.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000560430.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000560430.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000561282.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000561282.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000561282.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000561282.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000561282.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000561282.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000561282.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000561282.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000561282.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000561360.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000561360.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000561360.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000561360.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000561360.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000561360.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000561360.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000561360.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 ENST00000561360.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003940.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003942.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003942.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003942.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003942.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003942.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003942.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003942.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003942.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003943.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003943.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003943.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003943.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003943.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003943.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003943.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003943.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003943.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_001003943.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_033503.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_033503.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_033503.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_033503.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_033503.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_033503.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_033503.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_033503.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_033503.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009082.hg.1 NM_033503.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0