This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00373216_m1
See other ACMSD GE Assays ›
Gene Symbol:
ACMSD
Gene Name:
aminocarboxymuconate semialdehyde decarboxylase
Gene Aliases:
-
Chromosome Location:
Chr.2: 134838319 - 134902906 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00373216_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ACMSD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001307983.1 NP_001294912.1 1 - 2 126 118
GenBank mRNA BC016018.1 - 1 - 2 128 4714682 118

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200009424.hg.1

Additional Information:

This assay is in the 5'UTR region of the BC016018.1,NM_001307983.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Panther Classification:

Molecular Function -

decarboxylase

Gene Ontology Categories:

Function(s) Process(es)

tryptophan catabolic process
quinolinate metabolic process
protein oligomerization
regulation of quinolinate biosynthetic process
picolinic acid biosynthetic process
regulation of 'de novo' NAD biosynthetic process from tryptophan
aminocarboxymuconate-semialdehyde decarboxylase activity
protein binding
zinc ion binding
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000498093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000498093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000498093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 ENST00000498093.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay PSR0200165882.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200165882.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay PSR0200165882.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200165882.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay PSR0200165882.hg.1 TC0200009424.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000498093.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000498093.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000498093.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 ENST00000498093.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay TC0200009424.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay PSR0200165879.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200165879.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200165879.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200165879.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay PSR0200165879.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay PSR0200165879.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200165879.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200165879.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200165879.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay PSR0200165879.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay PSR0200165879.hg.1 TC0200009424.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200165881.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200165881.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200165881.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200165881.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay PSR0200165881.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay PSR0200165881.hg.1 ENST00000356140.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200165881.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200165881.hg.1 ENST00000485893.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200165881.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay PSR0200165881.hg.1 NM_138326.2 Refseq
Affymetrix Human Clariom D Assay PSR0200165881.hg.1 TC0200009424.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200165883.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200165883.hg.1 ENST00000498093.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200165883.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay PSR0200165883.hg.1 ENST00000392928.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200165883.hg.1 ENST00000498093.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200165883.hg.1 NM_001307983.1 Refseq
Affymetrix Human Clariom D Assay PSR0200165883.hg.1 TC0200009424.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0