This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00373422_m1
See other CD200R1 GE Assays ›
Gene Symbol:
CD200R1
Gene Name:
CD200 receptor 1
Gene Aliases:
CD200R, HCRTR2, MOX2R, OX2R
Chromosome Location:
Chr.3: 112922685 - 112975090 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00373422_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CD200R1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138806.3 NP_620161.1 1 - 2 302 105
NM_138939.2 NP_620385.1 1 - 2 302 105
GenBank mRNA AF495380.1 - 1 - 2 124 105
AF497549.1 - 1 - 2 124 105
AK293071.1 - 1 - 2 247 105
AY284975.1 - 1 - 2 291 105
AY358910.1 - 1 - 2 327 105
BC069661.1 - 1 - 2 100 7262527 105
BC069721.1 - 1 - 2 100 7262539 105
BC069743.1 - 1 - 2 100 7262551 105
BC143393.1 - 1 - 2 148 9051899 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300012007.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001654.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CD200R1

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

viral process
regulation of immune response
protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03029822.hg.1 ENST00000308611.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03029822.hg.1 ENST00000440122.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03029822.hg.1 NM_138806.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03029822.hg.1 NM_138939.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03029822.hg.1 TC03001654.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0300189404.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300189404.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300189404.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay PSR0300189404.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay PSR0300189404.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300189404.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300189404.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay PSR0300189404.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay PSR0300189404.hg.1 TC0300012007.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000295863.4 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000308611.7 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000440122.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000471858.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 ENST00000490004.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138806.3 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138939.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_138940.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Affymetrix Human Clariom D Assay TC0300012007.hg.1 NM_170780.2 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 106142 - -
ABI Human Genome Survey Microarray Version 2.0 177176 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline