This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00373748_m1
See other LIPH GE Assays ›
Gene Symbol:
LIPH
Gene Name:
lipase H
Gene Aliases:
AH, ARWH2, HYPT7, LAH2, LPDLR, PLA1B, mPA-PLA1
Chromosome Location:
Chr.3: 185506262 - 185552661 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00373748_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIPH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_139248.2 NP_640341.1 1 - 2 157 103
XM_006713529.3 XP_006713592.1 1 - 2 164 103
XM_017005852.1 XP_016861341.1 1 - 2 164 103
GenBank mRNA AK122651.1 - 1 - 2 157 103
AK298380.1 - 1 - 2 132 103
AY036912.1 - 1 - 2 137 103
AY093498.1 - 1 - 2 129 103
BC064941.1 - 1 - 2 144 6006910 103
EF186228.1 - 1 - 2 134 103
EF186229.1 - 1 - 2 134 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300013417.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03002078.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK122651.1,AK298380.1,AY036912.1,AY093498.1,BC064941.1,EF186228.1,EF186229.1,NM_139248.2,XM_006713529.3,XM_017005852.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LIPH

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid catabolic process
lipid digestion
phospholipase activity
heparin binding
carboxylic ester hydrolase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000429510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000429510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000429510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000429510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000435679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000435679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000435679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000435679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000452897.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 ENST00000452897.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03036805.hg.1 TC03002078.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03036805.hg.1 ENST00000296252.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03036805.hg.1 ENST00000424591.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03036805.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03036807.hg.1 TC03002078.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03036807.hg.1 ENST00000296252.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03036807.hg.1 ENST00000424591.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03036807.hg.1 NM_139248.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03036808.hg.1 TC03002078.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03036808.hg.1 ENST00000296252.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03036808.hg.1 ENST00000424591.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03036808.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000429510.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000429510.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000429510.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000429510.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000435679.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000435679.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000435679.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000435679.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000452897.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 ENST00000452897.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay TC0300013417.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay PSR0300200017.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay PSR0300200017.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300200017.hg.1 ENST00000429510.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300200017.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay PSR0300200017.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay PSR0300200017.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300200017.hg.1 ENST00000429510.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300200017.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay PSR0300200017.hg.1 TC0300013417.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300200020.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay PSR0300200020.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300200020.hg.1 ENST00000429510.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300200020.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay PSR0300200020.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay PSR0300200020.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300200020.hg.1 ENST00000429510.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300200020.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay PSR0300200020.hg.1 TC0300013417.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300200019.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay PSR0300200019.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300200019.hg.1 ENST00000429510.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300200019.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay PSR0300200019.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay PSR0300200019.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300200019.hg.1 ENST00000429510.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300200019.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay PSR0300200019.hg.1 TC0300013417.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300200021.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay PSR0300200021.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300200021.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay PSR0300200021.hg.1 ENST00000296252.8 Ensembl
Affymetrix Human Clariom D Assay PSR0300200021.hg.1 ENST00000424591.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300200021.hg.1 NM_139248.2 Refseq
Affymetrix Human Clariom D Assay PSR0300200021.hg.1 TC0300013417.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 186517 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0