| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02008449.hg.1 |
ENST00000283646.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02008449.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02008449.hg.1 |
TC02000546.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200158059.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200158059.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200158059.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200158059.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200158059.hg.1 |
TC0200008351.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
ENST00000283646.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008351.hg.1 |
NM_144563.2 |
Refseq |