This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00375898_m1
See other RPIA GE Assays ›
Gene Symbol:
RPIA
Gene Name:
ribose 5-phosphate isomerase A
Gene Aliases:
RPI, RPIAD
Chromosome Location:
Chr.2: 88691658 - 88750935 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00375898_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 190.28
190.28
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPIA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144563.2 NP_653164.2 4 - 5 500 67
GenBank mRNA AY050633.1 - 4 - 5 476 67
BC015529.1 - 4 - 5 472 3904718 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200008351.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02000546.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RPIA

Panther Classification:

Molecular Function -

isomerase

Gene Ontology Categories:

Function(s) Process(es)

pentose-phosphate shunt
pentose-phosphate shunt, non-oxidative branch
ribose-5-phosphate isomerase activity
protein binding
monosaccharide binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02008451.hg.1 ENST00000283646.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02008451.hg.1 ENST00000543560.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02008451.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02008451.hg.1 TC02000546.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0200158061.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200158061.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay PSR0200158061.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200158061.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay PSR0200158061.hg.1 TC0200008351.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-5xwv5:80/100.66.76.242:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0