This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00376786_m1
See other PACRGL GE Assays ›
Gene Symbol:
PACRGL
Gene Name:
PARK2 coregulated like
Gene Aliases:
C4orf28
Chromosome Location:
Chr.4: 20696282 - 20757766 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00376786_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PACRGL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130727.1 NP_001124199.1 4 - 5 704 100
NM_001317849.1 NP_001304778.1 5 - 6 799 100
NM_145048.3 NP_659485.1 7 - 8 998 100
XM_011513805.2 XP_011512107.1 7 - 8 783 100
XM_017007747.1 XP_016863236.1 9 - 10 997 100
XM_017007751.1 XP_016863240.1 8 - 9 1065 100
XM_017007752.1 XP_016863241.1 7 - 8 912 100
XM_017007753.1 XP_016863242.1 7 - 8 782 100
XM_017007754.1 XP_016863243.1 5 - 6 839 100
XM_017007755.1 XP_016863244.1 5 - 6 656 100
XM_017007756.1 XP_016863245.1 5 - 6 649 100
XM_017007757.1 XP_016863246.1 5 - 6 585 100
XM_017007758.1 XP_016863247.1 4 - 5 534 100
XM_017007759.1 XP_016863248.1 4 - 5 482 100
GenBank mRNA AK297556.1 - 4 - 5 515 100
BC023002.2 - 7 - 8 745 4992139 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000161.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PACRGL

Gene Ontology Categories:

Process(es)

protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR04002574.hg.1 ENST00000295290.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002574.hg.1 ENST00000360916.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002574.hg.1 ENST00000444671.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002574.hg.1 ENST00000502374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002574.hg.1 ENST00000502938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002574.hg.1 ENST00000503585.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002574.hg.1 ENST00000513459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002574.hg.1 ENST00000538990.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002574.hg.1 NM_001130727.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04002574.hg.1 NM_145048.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04002574.hg.1 TC04000161.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04002573.hg.1 ENST00000295290.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002573.hg.1 ENST00000360916.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002573.hg.1 ENST00000444671.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002573.hg.1 ENST00000502374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002573.hg.1 ENST00000502938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002573.hg.1 ENST00000503585.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002573.hg.1 ENST00000513459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002573.hg.1 ENST00000538990.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002573.hg.1 NM_001130727.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04002573.hg.1 NM_145048.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04002573.hg.1 TC04000161.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04002576.hg.1 ENST00000295290.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002576.hg.1 ENST00000360916.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002576.hg.1 ENST00000444671.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002576.hg.1 ENST00000503585.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002576.hg.1 ENST00000513459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002576.hg.1 ENST00000538990.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002576.hg.1 NM_001130727.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04002576.hg.1 NM_145048.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04002576.hg.1 TC04000161.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04002572.hg.1 ENST00000295290.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002572.hg.1 ENST00000360916.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002572.hg.1 ENST00000444671.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002572.hg.1 ENST00000502374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002572.hg.1 ENST00000502938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002572.hg.1 ENST00000503585.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002572.hg.1 ENST00000507634.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002572.hg.1 ENST00000513459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002572.hg.1 ENST00000538990.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002572.hg.1 NM_001130727.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04002572.hg.1 NM_145048.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04002572.hg.1 TC04000161.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04002580.hg.1 ENST00000295290.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002580.hg.1 ENST00000360916.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002580.hg.1 ENST00000444671.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002580.hg.1 ENST00000513459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002580.hg.1 NM_001130727.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04002580.hg.1 NM_145048.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04002580.hg.1 TC04000161.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04002575.hg.1 ENST00000295290.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002575.hg.1 ENST00000360916.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002575.hg.1 ENST00000444671.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002575.hg.1 ENST00000502374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002575.hg.1 ENST00000503585.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002575.hg.1 ENST00000513459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002575.hg.1 ENST00000538990.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002575.hg.1 NM_001130727.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04002575.hg.1 NM_145048.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04002575.hg.1 TC04000161.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04002578.hg.1 ENST00000295290.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002578.hg.1 ENST00000360916.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002578.hg.1 ENST00000444671.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002578.hg.1 ENST00000503585.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002578.hg.1 ENST00000513459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002578.hg.1 ENST00000538990.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04002578.hg.1 NM_001130727.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04002578.hg.1 NM_145048.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04002578.hg.1 TC04000161.hg.1 HTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline