This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00378112_m1
See other EEF1AKMT1 GE Assays ›
Gene Symbol:
EEF1AKMT1
Gene Name:
eukaryotic translation elongation factor 1 alpha lysine methyltransferase 1
Gene Aliases:
ESP13, N6AMT2
Chromosome Location:
Chr.13: 20728931 - 20773958 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00378112_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EEF1AKMT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001318939.1 NP_001305868.1 2 - 3 199 90
NM_174928.2 NP_777588.1 2 - 3 263 90
XM_017020432.1 XP_016875921.1 2 - 3 298 90
GenBank mRNA AK316604.1 - 2 - 3 197 90
BC018091.1 - 2 - 3 223 4793121 90
DQ823637.1 - 2 - 3 228 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300008264.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000460.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EEF1AKMT1

Gene Ontology Categories:

Function(s) Process(es)

peptidyl-lysine methylation
nucleic acid binding
protein-lysine N-methyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR13005239.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005239.hg.1 NM_174928.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13005239.hg.1 TC13000460.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR13005235.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005235.hg.1 ENST00000382758.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005235.hg.1 ENST00000473438.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005235.hg.1 NM_174928.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13005235.hg.1 TC13000460.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000460374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000460374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 ENST00000460374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 NM_174928.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 NM_174928.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 NM_174928.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 NM_174928.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 NM_174928.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008264.hg.1 NM_174928.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13005236.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005236.hg.1 ENST00000382758.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005236.hg.1 NM_174928.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13005236.hg.1 TC13000460.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR13005238.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005238.hg.1 ENST00000382758.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005238.hg.1 ENST00000473438.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13005238.hg.1 NM_174928.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13005238.hg.1 TC13000460.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1300155786.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300155786.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300155786.hg.1 ENST00000460374.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300155786.hg.1 NM_174928.2 Refseq
Affymetrix Human Clariom D Assay PSR1300155786.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300155786.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300155786.hg.1 ENST00000460374.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300155786.hg.1 NM_174928.2 Refseq
Affymetrix Human Clariom D Assay PSR1300155786.hg.1 TC1300008264.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1300155791.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300155791.hg.1 NM_174928.2 Refseq
Affymetrix Human Clariom D Assay PSR1300155791.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300155791.hg.1 NM_174928.2 Refseq
Affymetrix Human Clariom D Assay PSR1300155791.hg.1 TC1300008264.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1300155784.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300155784.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300155784.hg.1 NM_174928.2 Refseq
Affymetrix Human Clariom D Assay PSR1300155784.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Clariom D Assay PSR1300155784.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300155784.hg.1 NM_174928.2 Refseq
Affymetrix Human Clariom D Assay PSR1300155784.hg.1 TC1300008264.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382754.4 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000382758.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000460374.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000460374.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 ENST00000460374.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008264.hg.1 NM_174928.2 Refseq
Affymetrix Human Clariom D Assay TC1300008264.hg.1 NM_174928.2 Refseq
Affymetrix Human Clariom D Assay TC1300008264.hg.1 NM_174928.2 Refseq
Affymetrix Human Clariom D Assay TC1300008264.hg.1 NM_174928.2 Refseq
Affymetrix Human Clariom D Assay TC1300008264.hg.1 NM_174928.2 Refseq
Affymetrix Human Clariom D Assay TC1300008264.hg.1 NM_174928.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline