This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00378398_g1
See other ERAL1 GE Assays ›
Gene Symbol:
ERAL1
Gene Name:
Era like 12S mitochondrial rRNA chaperone 1
Gene Aliases:
CEGA, ERA, ERA-W, ERAL1A, H-ERA, HERA-A, HERA-B
Chromosome Location:
Chr.17: 28854957 - 28861067 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00378398_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ERAL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001317985.1 NP_001304914.1 6 - 7 783 79
NM_005702.3 NP_005693.1 6 - 7 786 79
NR_134328.1 6 - 7 786 79
GenBank mRNA AB049388.1 - 6 - 7 734 79
AF082657.1 - 6 - 7 727 79
AF082658.1 - 6 - 7 727 79
AK023342.1 - 6 - 7 727 79
AY007435.1 - 6 - 7 729 79
BC019094.2 - 6 - 7 718 4893068 79
BX384612.2 - 6 - 7 705 79
BX537542.1 - 5 - 6 1655 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700007399.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17000332.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ERAL1

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

ribosomal small subunit assembly
GTPase activity
protein binding
GTP binding
rRNA binding
ribosomal small subunit binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR17004112.hg.1 ENST00000254928.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17004112.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17004112.hg.1 TC17000332.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000578001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000578001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000578001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000578001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000583487.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000583487.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000583487.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 ENST00000583487.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay PSR1700152742.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay PSR1700152742.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Clariom D Assay PSR1700152742.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700152742.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700152742.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay PSR1700152742.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay PSR1700152742.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Clariom D Assay PSR1700152742.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700152742.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700152742.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay PSR1700152742.hg.1 TC1700007399.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000254928.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000412138.6 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000461894.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000471992.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000577942.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000578001.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000578001.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000578001.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000578001.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000580917.2 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000583487.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000583487.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000583487.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 ENST00000583487.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq
Affymetrix Human Clariom D Assay TC1700007399.hg.1 NM_005702.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-7669c5b596-474fd:80/100.66.77.40:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0