This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00379179_m1
See other METTL6 GE Assays ›
Gene Symbol:
METTL6
Gene Name:
methyltransferase like 6
Gene Aliases:
-
Chromosome Location:
Chr.3: 15381275 - 15427580 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00379179_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: METTL6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301791.1 NP_001288720.1 2 - 3 493 72
NM_001301792.1 NP_001288721.1 2 - 3 521 72
NM_152396.3 NP_689609.2 2 - 3 493 72
XM_005264867.3 XP_005264924.1 2 - 3 493 72
XM_005264870.4 XP_005264927.1 2 - 3 493 72
XM_006712970.3 XP_006713033.1 2 - 3 369 72
XM_006712972.3 XP_006713035.1 2 - 3 493 72
XM_011533357.2 XP_011531659.1 2 - 3 493 72
XM_017005718.1 XP_016861207.1 2 - 3 369 72
XM_017005722.1 XP_016861211.1 2 - 3 493 72
XM_017005724.1 XP_016861213.1 2 - 3 493 72
GenBank mRNA BC022400.1 - 2 - 3 450 4693314 72
BG701912.1 - 2 - 3 495 72
BI913767.1 - 2 - 3 488 72
BU183521.1 - 2 - 3 431 72

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene METTL6

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

methylation
methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline