This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00379287_m1
See other SLC7A14 GE Assays ›
Gene Symbol:
SLC7A14
Gene Name:
solute carrier family 7 member 14
Gene Aliases:
PPP1R142
Chromosome Location:
Chr.3: 170459554 - 170586074 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00379287_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC7A14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020949.2 NP_066000.2 5 - 6 1222 117
GenBank mRNA AB046833.1 - 4 - 5 673 117
AK122655.1 - 5 - 6 1217 117
BC022968.1 - 4 - 5 838 117

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300013095.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001991.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC7A14

Panther Classification:

Molecular Function -

amino acid transporter

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of phosphatase activity
L-amino acid transport
L-alpha-amino acid transmembrane transport
L-amino acid transmembrane transporter activity
antiporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03035623.hg.1 TC03001991.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03035623.hg.1 ENST00000231706.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03035623.hg.1 NM_020949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 ENST00000466168.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 ENST00000466168.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03035624.hg.1 TC03001991.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03035624.hg.1 ENST00000231706.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03035624.hg.1 NM_020949.2 Refseq
Affymetrix Human Clariom D Assay TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013095.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013095.hg.1 ENST00000466168.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013095.hg.1 ENST00000466168.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Clariom D Assay TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Clariom D Assay TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Clariom D Assay TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Clariom D Assay TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Clariom D Assay TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Clariom D Assay TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Clariom D Assay TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Clariom D Assay TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Clariom D Assay TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Clariom D Assay TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Clariom D Assay TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Clariom D Assay TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Clariom D Assay TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Clariom D Assay TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Clariom D Assay TC0300013095.hg.1 NM_020949.2 Refseq
Affymetrix Human Clariom D Assay PSR0300197952.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300197952.hg.1 NM_020949.2 Refseq
Affymetrix Human Clariom D Assay PSR0300197952.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300197952.hg.1 NM_020949.2 Refseq
Affymetrix Human Clariom D Assay PSR0300197952.hg.1 TC0300013095.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300197951.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300197951.hg.1 NM_020949.2 Refseq
Affymetrix Human Clariom D Assay PSR0300197951.hg.1 ENST00000231706.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300197951.hg.1 NM_020949.2 Refseq
Affymetrix Human Clariom D Assay PSR0300197951.hg.1 TC0300013095.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gqkn5:80/100.66.76.113:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0