This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00382661_m1
See other ESD GE Assays ›
Gene Symbol:
ESD
Gene Name:
esterase D
Gene Aliases:
FGH
Chromosome Location:
Chr.13: 46771256 - 46797681 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00382661_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ESD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001984.1 NP_001975.1 1 - 2 125 90
XM_017020414.1 XP_016875903.1 1 - 2 30 90
GenBank mRNA AF112219.1 - 1 - 2 125 90
AK098540.1 - 1 - 2 54 90
BC001169.1 - 1 - 2 40 3528593 90
D28416.1 - 1 - 2 41 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300008855.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000647.hg.1

Additional Information:

This assay is in the 5'UTR region of the AF112219.1,BC001169.1,D28416.1,NM_001984.1,XM_017020414.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ESD

Panther Classification:

Molecular Function -

protease serine protease protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

biological_process
formaldehyde catabolic process
glutathione derivative biosynthetic process
protein binding
hydrolase activity, acting on ester bonds
S-formylglutathione hydrolase activity
methylumbelliferyl-acetate deacetylase activity
carboxylic ester hydrolase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR13007390.hg.1 ENST00000378697.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13007390.hg.1 ENST00000378720.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13007390.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13007390.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13007390.hg.1 TC13000647.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR13007385.hg.1 ENST00000378697.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13007385.hg.1 ENST00000378720.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13007385.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13007385.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13007385.hg.1 TC13000647.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR13007389.hg.1 ENST00000378697.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13007389.hg.1 ENST00000378720.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13007389.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13007389.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13007389.hg.1 TC13000647.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR13007394.hg.1 ENST00000378720.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13007394.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13007394.hg.1 TC13000647.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000471867.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000471867.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000471867.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000471867.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000471867.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13007387.hg.1 ENST00000378697.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13007387.hg.1 ENST00000378720.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13007387.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13007387.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13007387.hg.1 TC13000647.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR13007392.hg.1 ENST00000378697.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13007392.hg.1 ENST00000378720.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13007392.hg.1 NM_001984.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13007392.hg.1 TC13000647.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1300159800.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300159800.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay PSR1300159800.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300159800.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay PSR1300159800.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300159800.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay PSR1300159800.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300159800.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay PSR1300159800.hg.1 TC1300008855.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1300159798.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300159798.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay PSR1300159798.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300159798.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300159798.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay PSR1300159798.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300159798.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay PSR1300159798.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300159798.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300159798.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay PSR1300159798.hg.1 TC1300008855.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1300159805.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300159805.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay PSR1300159805.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300159805.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay PSR1300159805.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300159805.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay PSR1300159805.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300159805.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay PSR1300159805.hg.1 TC1300008855.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1300159802.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300159802.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay PSR1300159802.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300159802.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay PSR1300159802.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300159802.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay PSR1300159802.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300159802.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay PSR1300159802.hg.1 TC1300008855.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1300159808.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300159808.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay PSR1300159808.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay PSR1300159808.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300159808.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay PSR1300159808.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay PSR1300159808.hg.1 TC1300008855.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378697.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000412582.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000471867.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000471867.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000471867.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000471867.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000471867.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 ENST00000495654.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay TC1300008855.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay PSR1300159811.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay PSR1300159811.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay PSR1300159811.hg.1 ENST00000378720.7 Ensembl
Affymetrix Human Clariom D Assay PSR1300159811.hg.1 NM_001984.1 Refseq
Affymetrix Human Clariom D Assay PSR1300159811.hg.1 TC1300008855.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 165885 - -
ABI Human Genome Survey Microarray Version 2.0 232746 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0