This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00382744_m1
See other IFIT3 GE Assays ›
Gene Symbol:
IFIT3
Gene Name:
interferon induced protein with tetratricopeptide repeats 3
Gene Aliases:
CIG-49, GARG-49, IFI60, IFIT4, IRG2, ISG60, P60, RIG-G, cig41
Chromosome Location:
Chr.10: 89327819 - 89340968 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00382744_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IFIT3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001031683.3 NP_001026853.1 1 - 2 146 91
GenBank mRNA AK297137.1 - 1 - 2 133 91
BC001383.2 - 1 - 2 79 3050764 91
BC004977.1 - 1 - 2 52 2906188 91
DB268497.1 - 1 - 2 146 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000008397.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000637.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK297137.1,BC001383.2,BC004977.1,NM_001031683.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IFIT3

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cell proliferation
response to virus
cellular response to interferon-alpha
negative regulation of apoptotic process
defense response to virus
type I interferon signaling pathway
molecular_function
RNA binding
protein binding
identical protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10007024.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10007024.hg.1 ENST00000371818.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10007024.hg.1 ENST00000543062.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10007024.hg.1 NM_001031683.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10007024.hg.1 NM_001549.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10007024.hg.1 TC10000637.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10007023.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10007023.hg.1 ENST00000543062.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10007023.hg.1 NM_001031683.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10007023.hg.1 TC10000637.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10007022.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10007022.hg.1 ENST00000543062.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10007022.hg.1 NM_001031683.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10007022.hg.1 TC10000637.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 ENST00000371818.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 ENST00000371818.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 ENST00000371818.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 ENST00000371818.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001031683.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001031683.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001031683.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001031683.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001031683.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001031683.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001289758.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001289758.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001289758.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001289758.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001289758.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001289758.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001289759.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001289759.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001289759.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001289759.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001289759.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001289759.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001289759.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001549.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001549.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001549.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001549.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008397.hg.1 NM_001549.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10007021.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10007021.hg.1 ENST00000543062.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10007021.hg.1 NM_001031683.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10007021.hg.1 TC10000637.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1000156932.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000156932.hg.1 ENST00000371818.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000156932.hg.1 NM_001031683.3 Refseq
Affymetrix Human Clariom D Assay PSR1000156932.hg.1 NM_001289758.1 Refseq
Affymetrix Human Clariom D Assay PSR1000156932.hg.1 NM_001289759.1 Refseq
Affymetrix Human Clariom D Assay PSR1000156932.hg.1 NM_001549.5 Refseq
Affymetrix Human Clariom D Assay PSR1000156932.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000156932.hg.1 ENST00000371818.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000156932.hg.1 NM_001031683.3 Refseq
Affymetrix Human Clariom D Assay PSR1000156932.hg.1 NM_001289758.1 Refseq
Affymetrix Human Clariom D Assay PSR1000156932.hg.1 NM_001289759.1 Refseq
Affymetrix Human Clariom D Assay PSR1000156932.hg.1 NM_001549.5 Refseq
Affymetrix Human Clariom D Assay PSR1000156932.hg.1 TC1000008397.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000008397.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008397.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008397.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008397.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008397.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008397.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Clariom D Assay TC1000008397.hg.1 ENST00000371818.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008397.hg.1 ENST00000371818.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008397.hg.1 ENST00000371818.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008397.hg.1 ENST00000371818.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001031683.3 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001031683.3 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001031683.3 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001031683.3 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001031683.3 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001031683.3 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001289758.1 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001289758.1 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001289758.1 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001289758.1 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001289758.1 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001289758.1 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001289759.1 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001289759.1 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001289759.1 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001289759.1 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001289759.1 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001289759.1 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001289759.1 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001549.5 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001549.5 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001549.5 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001549.5 Refseq
Affymetrix Human Clariom D Assay TC1000008397.hg.1 NM_001549.5 Refseq
Affymetrix Human Clariom D Assay PSR1000156927.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000156927.hg.1 NM_001031683.3 Refseq
Affymetrix Human Clariom D Assay PSR1000156927.hg.1 NM_001289759.1 Refseq
Affymetrix Human Clariom D Assay PSR1000156927.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000156927.hg.1 NM_001031683.3 Refseq
Affymetrix Human Clariom D Assay PSR1000156927.hg.1 NM_001289759.1 Refseq
Affymetrix Human Clariom D Assay PSR1000156927.hg.1 TC1000008397.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1000156926.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000156926.hg.1 NM_001031683.3 Refseq
Affymetrix Human Clariom D Assay PSR1000156926.hg.1 NM_001289759.1 Refseq
Affymetrix Human Clariom D Assay PSR1000156926.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000156926.hg.1 NM_001031683.3 Refseq
Affymetrix Human Clariom D Assay PSR1000156926.hg.1 NM_001289759.1 Refseq
Affymetrix Human Clariom D Assay PSR1000156926.hg.1 TC1000008397.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1000156928.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000156928.hg.1 NM_001031683.3 Refseq
Affymetrix Human Clariom D Assay PSR1000156928.hg.1 NM_001289759.1 Refseq
Affymetrix Human Clariom D Assay PSR1000156928.hg.1 ENST00000371811.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000156928.hg.1 NM_001031683.3 Refseq
Affymetrix Human Clariom D Assay PSR1000156928.hg.1 NM_001289759.1 Refseq
Affymetrix Human Clariom D Assay PSR1000156928.hg.1 TC1000008397.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 202937 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0