This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00383058_m1
See other GJB5 GE Assays ›
Gene Symbol:
GJB5
Gene Name:
gap junction protein beta 5
Gene Aliases:
CX31.1
Chromosome Location:
Chr.1: 34754777 - 34758513 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00383058_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GJB5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005268.3 NP_005259.1 1 - 2 149 110
GenBank mRNA AF052693.1 - 1 - 2 143 110
AK129509.1 - 1 - 2 149 110
BC004379.1 - 1 - 2 76 3639452 110

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF052693.1, BC004379.1, NM_005268.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GJB5

Panther Classification:

Molecular Function -

gap junction

Gene Ontology Categories:

Function(s)

cell communication
epidermis development
trophoblast giant cell differentiation
spongiotrophoblast differentiation
labyrinthine layer morphogenesis

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100007745.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007745.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007745.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007745.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007745.hg.1 NM_005268.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007745.hg.1 NM_005268.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007745.hg.1 NM_005268.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007745.hg.1 NM_005268.3 Refseq
Affymetrix Human Clariom D Assay TC0100007745.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007745.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007745.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007745.hg.1 ENST00000338513.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007745.hg.1 NM_005268.3 Refseq
Affymetrix Human Clariom D Assay TC0100007745.hg.1 NM_005268.3 Refseq
Affymetrix Human Clariom D Assay TC0100007745.hg.1 NM_005268.3 Refseq
Affymetrix Human Clariom D Assay TC0100007745.hg.1 NM_005268.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline